Search
Search

atp2a1| Gene Symbol: atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032327.1 | NP_001027498.1 | 1 - 2 | 296 | 81 | |
| XM_012954931.2 | XP_012810385.1 | 1 - 2 | 328 | 81 | ||
| GenBank mRNA | BC096519.1 | - | 1 - 2 | 296 | 81 | |
Set Membership: |
|