Search
Search

kif11| Gene Symbol: kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016116.3 | NP_001016116.2 | 7 - 8 | 870 | 79 | |
| XM_012966561.1 | XP_012822015.1 | 8 - 9 | 1030 | 79 | ||
| GenBank mRNA | BC166147.1 | - | 7 - 8 | 801 | 79 | |
| CR762094.2 | - | 7 - 8 | 870 | 79 | ||
| EF524557.1 | - | 6 - 7 | 689 | 79 | ||
Set Membership: |
|