Search
Search

clvs1| Gene Symbol: clvs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097346.1 | NP_001090815.1 | 4 - 5 | 962 | 97 | |
| XM_012964142.2 | XP_012819596.1 | 4 - 5 | 977 | 97 | ||
| GenBank mRNA | BC135266.1 | - | 4 - 5 | 962 | 97 | |
Set Membership: |
|