Search
Search

enosf1| Gene Symbol: enosf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113128.1 | NP_001106599.1 | 14 - 15 | 1129 | 105 | |
| XM_012964234.2 | XP_012819688.1 | 15 - 16 | 1559 | 105 | ||
| XM_012964235.2 | XP_012819689.1 | 15 - 16 | 1476 | 105 | ||
| XM_012964236.2 | XP_012819690.1 | 15 - 16 | 1450 | 105 | ||
| XM_018094523.1 | XP_017950012.1 | 14 - 15 | 1648 | 105 | ||
| GenBank mRNA | BC155430.1 | - | 14 - 15 | 1129 | 105 | |
Set Membership: |
|