Search
Search

eln2| Gene Symbol: eln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142826.2 | NP_001136298.1 | 38 - 39 | 1701 | 77 | |
| XM_012970193.2 | XP_012825647.1 | 37 - 38 | 1694 | 77 | ||
| XM_012970194.2 | XP_012825648.1 | 37 - 38 | 1679 | 77 | ||
| XM_012970196.2 | XP_012825650.1 | 35 - 36 | 1610 | 77 | ||
| XM_012970197.2 | XP_012825651.1 | 35 - 36 | 1610 | 77 | ||
| GenBank mRNA | CU075851.1 | - | 38 - 39 | 1854 | 77 | |
| EU220008.1 | - | 38 - 39 | 1703 | 77 | ||
Set Membership: |
|