Search
Search

gst2| Gene Symbol: gst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111896.1 | NP_001105366.1 | 305 | 77 | ||
| GenBank mRNA | AY103712.2 | - | 316 | 77 | ||
| BT063667.1 | - | 309 | 77 | |||
| U12679.1 | - | 305 | 77 | |||
| X79515.1 | - | 306 | 77 | |||
Set Membership: |
|