Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492679_m1
          See other IRF9 GE Assays ›
          Gene Symbol:
          Irf9
          Gene Name:
          interferon regulatory factor 9
          Gene Aliases:
          Irf-9, Isgf3g, p48
          Chromosome Location:
          Chr.14: 55603985 - 55610030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492679_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irf9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159417.1 NP_001152889.1 8 - 9 1326 91
          NM_001159418.1 NP_001152890.1 7 - 8 1124 91
          NM_008394.3 NP_032420.1 7 - 8 1240 91
          GenBank mRNA AK145911.1 - 6 - 7 992 91
          BC005435.1 - 8 - 9 1101 3601255 91
          BC012968.1 - 7 - 8 1135 3257714 91
          U51992.1 - 7 - 8 1044 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002834.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irf9

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          type I interferon biosynthetic process
          regulatory region DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114938 BC012968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114938 BC005435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114938 AK145911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114938 NM_001159418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114938 NM_008394.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146488 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136413 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114938 BC012968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114938 BC005435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114938 AK145911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114938 NM_001159418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114938 NM_008394.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P176013 BC012968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P176013 CK788642.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P176013 U51992.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P176013 AK149742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P176013 BC005435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127367 BC012968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127367 U51992.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127367 AK149742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127367 BC005435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127367 AK043697.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29618801 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08118961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08118971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136413 - -
          Whole Mouse Genome G4122A A_52_P176013 BC012968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P176013 CK788642.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P176013 U51992.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P176013 AK149742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P176013 BC005435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127367 BC012968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127367 U51992.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127367 AK149742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127367 BC005435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127367 AK043697.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130883 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136413 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11457298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11457297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11457289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11457299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29618788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11338869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11333706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11691484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11457283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11457289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11691490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11333705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29618818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415324 - -
          Affymetrix HT MG-430 PM Array Plate 1421322_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415319 - -
          Mouse Genome 430 2.0 Array 1421322_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421322_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16391_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16391_at - -
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_001159417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_001159417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_001159418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_001159418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_008394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 NM_008394.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006362.mm.1 TC1400002834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_001159417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_001159417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_001159418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_001159418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_008394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 NM_008394.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006363.mm.1 TC1400002834.mm.1 MTA TC
          Mouse Expression Set 430 1421322_a_at - -
          Mouse Expression Set 430 1421322_PM_a_at - -
          Murine Genome U74v2 Set 103634_PM_at - -
          Murine Genome U74v2 Set 103634_at - -
          Mouse Genome 430A 2.0 Array 1421322_a_at - -
          Mouse Genome 430A 2.0 Array 1421322_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421322_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421322_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919070 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3190131 - -
          Illumina mouse-6 v1.0 expression beadchip 3190131 - -
          Illumina mouse-6 v1.0 expression beadchip 7100142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636339 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233461 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636339 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233461 - -
          mouse-6 v1.1 (Illumina) scl000304.1_6-S - -
          mouse-6 v1.1 (Illumina) scl016391.8_62-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100142 - -
          Illumina Sentrix mouse-6.v1 bead array 3190131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016391.8_62-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000304.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190131 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636339 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233461 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636339 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233461 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl016391.8_62-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000304.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline