Search
Cancer biomarkers are molecules that can be detected in tumor tissue or blood (sometimes called liquid biopsy) that provide information about a tumor’s biology. These tumor-specific molecules can involve changes in the DNA and RNA or even changes at the protein level that may indicate sensitivity to a particular treatment (i.e., targeted therapy or immunotherapy). Biomarkers may enable precision oncology research by elucidating the biological mechanisms that drive tumor development, progression, and resistance.
Next-generation sequencing (NGS) is a powerful technology that allows for the simultaneous analysis of multiple biomarkers from a single sample in a cost- and time-efficient manner. While tissue NGS is the gold standard for gaining insights into the biology of a tumor, liquid biopsy–based NGS is emerging as a highly sensitive method to understand resistance mechanisms as the tumor evolves. Liquid biopsies samples may be used as an alternative when tissue samples are not available. Moreover, liquid biopsy testing may complement tissue testing to better understand inter- and intra-tumor heterogeneity.
Figure 1. Relevant and emerging biomarkers in different cancer types detected by Oncomine CGP research assays.
Recent advances in breast cancer research have uncovered many relevant and emerging biomarkers that go beyond immunohistochemical-based biomarkers. Find out why NGS technologies for broad molecular profiling are becoming necessary.
Figure 2. NGS uses a single assay to assess relevant and emerging biomarkers in breast cancer research.
BRCA1/2 proteins play critical roles in DNA repair by the homologous recombination repair (HRR) pathway, and loss of BRCA1/2 function has been well studied as a cause of homologous recombination deficiency (HRD). Learn how NGS can identify both causes and consequences of HRD.
Figure 3. HRR pathway. Non-gray genes are covered by the Oncomine Comprehensive Assay Plus. Teal genes were included in clinical trials of prostate cancer clinical research samples.
Estrogen Receptor 1 (ESR1) encodes estrogen receptor α and plays a critical role in breast cancer by regulating the response to estrogen. Mutations in ESR1 are an active area of investigation, and NGS allows for comprehensive analysis, unveiling the intricate genomic alterations and complex mechanisms.
Figure 4. Schematic diagram of ERα mutations and their frequencies in metastatic breast cancer after treatment.1
Activating mutations in KRAS are one of the most prevalent oncogenic drivers in non-small cell lung cancer (NSCLC) and mutant KRAS remains challenging to target. Learn how NGS can simultaneously analyze relevant biomarkers and co-mutations essential to understanding the genomic landscape of mutant KRAS tumors.
Figure 5. Prevalence of oncogenic drivers in NSCLC and frequency of KRAS variants.2,3
Oncomine Solution automated workflows enable oncology biomarker detection to help meet your laboratory research needs, even at varying levels of NGS expertise.
Download our Biomarker Guides to learn all you need to know in a few minutes:
References
1. Ma CX, Reinert T, Chmielewska I, Ellis MJ (2015) Mechanisms of aromatase inhibitor resistance. Nat Rev Cancer 15(5):261−275.
2. Tan AC, Tan DSW (2022) Targeted therapies for lung cancer patients with oncogenic driver molecular alterations. J Clin Oncol 40(6):611−625.
3. Salem ME, El-Refai SM, Sha W et al. (2022) Landscape of KRAS(G12C), associated genomic alterations, and interrelation with immuno-oncology biomarkers in KRAS-mutated cancers. JCO Precis Oncol 6:e2100245.
CN: 65538