Key methods for structural insights
We offer integrated solutions and support from sample preparation to data analysis for multiple cryo-EM methods, including single particle cryo-EM, microcrystal electron diffraction (MicroED), and cryo-tomography.
Single particle cryo-EM
Single particle cryo-EM can determine structures of challenging proteins and other macromolecules, without the need for crystals, while providing insight into how these proteins function in a way that other methods simply cannot. Single particle analysis is an increasingly popular cryo-EM technique that allows you to investigate biomolecules at near- and atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
Tundra Cryo-TEM
Dedicated structural analysis solution designed to bring cryo-EM to your laboratory.
Glacios 3 Cryo-TEM
An accessible, automated solution for high-resolution structure determination at 200 kV.
Krios 5 Cryo-TEM
Atomic resolution cryo-EM instrument with enhanced productivity and compact design.
Cryo-electron tomography
Cryo-electron tomography (cryo-ET) provides 3D snapshots of proteins at work within their functional cellular environments, allowing users to see and understand how they, and other molecules, work together to carry out major processes in a cell. This is because cryo-ET delivers both structural information about individual proteins as well as their spatial arrangements within the cell, making it a truly unique technique. Cryo-ET has enormous potential for cell biology as it bridges the gap between light microscopy and near-atomic resolution techniques like single-particle analysis.
Aquilos 2 Cryo-FIB
Dedicated cryo-FIB for cellular cryo-electron tomography sample preparation
Hydra Bio Plasma-FIB
Versatile plasma-FIB for multi-application labs
Arctis Cryo-Plasma-FIB
Automated cryo-plasma-FIB for throughput and connectivity in the cryo-electron tomography workflow
Microcrystal electron diffraction
Microcrystal electron diffraction (MicroED) is a technique that allows fast, high-resolution structural determination of small molecules and proteins. Atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture. 
CETA-D Camera and MicroED Package
A complete solution for performing MicroED on Thermo Scientific Cryo-TEMs, including hardware and software needed for fast, high-resolution 3D structure determination.
Talos 12 TEM
Versatile TEM and STEM microscope for 2D and 3D visualization of beam sensitive samples and materials.
Glacios 3 Cryo-TEM
An accessible, automated solution for high-resolution structure determination at 200 kV.
Krios 5 Cryo-TEM
Atomic resolution cryo-EM instrument with enhanced productivity and compact design.
For Research Use Only. Not for use in diagnostic procedures.