Search
The CellInsight family of HCS/HCA platforms offers widefield, confocal, or brightfield automated imaging modes, all within one protocol. Engineered with speed in mind, real-time imaging and analysis can be conducted in parallel for the fastest time-to-data possible. Learn more about our unique features including publication-quality imaging, real-time analysis software, cell-painting research, and database management software below.
The CellInsight family of high-content screening (HCS)/high-content analysis (HCA) platforms are designed to provide automated imaging versatility and accelerated throughput for the fastest time-to-data possible. Choose from widefield, confocal, or brightfield automated imaging modes, all within one protocol. Our instruments provide up to 12 colors to choose from, from ultraviolet through near-infrared fluorescence and multicolor brightfield imaging to enable flexible imaging paradigms. Finally, our systems are engineered with speed in mind, enabling real-time imaging and analysis conducted in parallel for the fastest time-to-data possible.
Breast cancer tissue cells
CellInsight CX7 LED Pro. Human breast cancer histology slides were stained with H&E and KI67-associated DAB markers (imaged with red, green, and blue transmitted light respectively and a 20X objective lens and the 3-channel Histology bioapplication.
A549 cells
CellInsight CX7 LED. Stains: HCS CellMask Blue (blue), Invitrogen Alexa Fluor 488 Phalloidin (green), and Invitrogen Alexa Fluor 750 goat anti–mouse IgG secondary antibody (pink).
BPAE cells
CellInsight CX5. Stains: DAPI (blue), Alexa Fluor 568 phalloidin (red), and goat anti–mouse IgG (H+L) secondary antibody, Alexa Fluor 488 conjugate (green).
HeLa cells
CellInsight CX5. Stains: Hoechst 33342 (blue) and Alexa Fluor 680 phalloidin (yellow).
The Thermo Scientific HCS Studio Cell Analysis Software expertly balances flexibility and ease of use to enable meaningful data analysis from single cells through populations. Parallel imaging and analysis offers true real-time results, fast.
HCS Studio software allows backtracking of each individual cell or cellular event to minimize artifacts in the data. View the corresponding cell level cutout associated with each data point to ensure quality control of what you are actually measuring.
To accelerate assay development, every parameter can be evaluated for assay performance based on Z-prime statistical analysis. This allows you to select your top-performing assays as you scale your screening campaigns. HCS Studio software also features guided workflows with user-friendly visual feedback to facilitate easy assay protocol setup, image acquisition, analysis, and scanning.
The HCS Studio software for CellInsight CX7 LED Pro and CellInsight CX7 LZR Pro also feature the Thermo Scientific EurekaScan Finder software. This software significantly helps reduce your instrument runtime and analysis efforts by automating the detection and subsequent capture of events at higher magnification.
EurekaScan Finder software offers two different approaches based on the type of sample and user-stipulated criteria that will inform high-content screening.
The CX7 LZR Pro HCS platform is equipped with a laser-based autofocus and a powerful sCMOS camera to boost your experiment's assay window and reduce exposure time, providing amazing acquisition speed. Assay speed is further enhanced with true on‑the-fly phenotyping that collapses analysis and image capture into one parallel stream, allowing you to get your data faster. Additionally, the new Thermo Scientific Image-iT™ Cell Painting Kit has been codeveloped with the CellInsight CX7 LZR Pro platform to enable turnkey multiparameter labeling, imaging, and analysis.
All these advanced features—paired with a dedicated, all-new, cell-painting bio-application in Thermo Scientific HCS Studio Software and optional Thermo Scientific Amira Software allow the CellInsight CX7 LZR Pro HCS Platform to simplify your most challenging assays, such as cell painting, morphological tracing in 3D, and immune cell to tumor colocalizations.
Thermo Scientific Store™ Image and Database Management Software is a SQL™ or Oracle™ relational database used for storing and managing the images and data automatically transferred from an HCS instrument or imported from another imaging source. It allows for rapid, highly scalable, global access to both data and images. Use the Store Image and Database Management Software after you have collected data and images.
Every Thermo Scientific High Content Platform is configured with a base version of Thermo Scientific Store Express Image and Database Management Software. With 10 GB of storage, Store Express Image and Database Management Software helps ensure the security of your data and allows you to be productive from your first day of use. Although it is not a scalable solution, you always have the option to upgrade to the full solution, Thermo Scientific Store Image and Database Management Software as your requirements change.
Store Image and Database Management Software’s optional configuration includes:
Store Image and Database Management Software expands to a server-based platform, allowing huge amounts of images and data to be managed. With Store Image and Database Management Software, import, convert and spool images from any platform or client running HCS Studio Cell Analysis Software. Share data and images across your organization for a fully collaborative work environment.
Product |
Quantity |
Cat. No. |
|
Thermo Scientific Store Informatics Software Standard Edition |
1 each |
S0100015D |
|
Thermo Scientific Store Informatics Software Academic Edition |
1 each |
S0100015A |
|
Thermo Scientific Store Informatics Software Enterprise Edition |
1 each |
S0100015 |
For Research Use Only. Not for use in diagnostic procedures.