Visibly simple food pathogen and quality PCR testing - simple workflows designed to get results
Rapidly and accurately detect relevant foodborne pathogens and other important food safety and quality targets with the Thermo Scientific SureTect Food Safety PCR System.  
Empower your operations with simple, scalable workflows and rapid reporting to enable faster product release and interventions. Our food safety PCR assays offer detection of the major pathogens and other important targets in food, beverage, pet food and animal feed, primary production samples, and environmental samples.
Whether your laboratory is looking to introduce PCR technology for the first time, or you want to expand the PCR testing services you offer, we can help you find the right solution.  Benefit from:
Case study: Bringing the benefits of PCR and automation to high-throughput testing with the SureTect Automation Platform
Streamlined workflow for reliable results
Single enrichment step
8-24 hours
Single enrichment step of 8-24 hours (depending on target and matrix)
Sample preparation and lysis
20-40 minutes
Fully automated using the SureTect Automation Workflow or manual preparation with direct lysis in the Applied Biosystems SimpliAmp Thermal Cycler
Run, read, and report
~80 minutes
Perform PCR with the Applied Biosystems QuantStudio 5 Instrument using the RapidFinder Analysis Software. Report PCR-negative results immediately (results are automatically interpreted)
Confirm PCR positives
24 hours
Confirm PCR-positive results, after overnight incubation, using rapid chromogenic culture media methods
SureTect Features
Streamlined enrichment
Single versus dual enrichment protocols for faster time-to-result and streamlined workflow
Validated assays
A range of assays validated for a broad range of targets and matrices according to certification requirements of AOAC and ISO 16140-2:2016 (by AFNOR or MicroVal)
Flexible setup
Universal PCR conditions for detecting multiple targets in the same run
Automatable workflows
Fully automated lysis and PCR setup with the Thermo Scientific SureTect Automation Platform
Intuitive instrumentation
‘Plug and play’ ready-to-use instruments out of the box for rapid pathogen dection
Smart software
Fully automated amplification, detection, data collection, and data analysis using Thermo Scientific RapidFinder Analysis Software v3.0 or later
Advancing poultry safety with confidence: Whitbeck Labs on SureTect PCR
Discover how Whitbeck Labs in Springdale, Arkansas leverages the Thermo Scientific SureTect PCR System to deliver fast, accurate, and reliable pathogen detection for the poultry industry and beyond. Hear firsthand from President and Microbiologist Gordon Whitbeck and his staff as they share how the SureTect System supports their USDA NPIP Salmonella testing, regulatory compliance, and commitment to high-quality microbiological services.
The SureTect PCR System – OMIC USA’s choice for reliable, robust foodborne pathogen detection 
After reviewing several PCR solutions for foodborne pathogen testing, OMIC selected the Thermo Scientific SureTect PCR System for its laboratories' pathogen testing, favoring its robust, efficient workflow, compatibility with a wide range of matrices, and strong performance during validation.
Visible selective enrichment of Salmonella and Cronobacter species
In your busy Salmonella and Cronobacter testing workflows, there is no time for error. A common mistake, particularly in high throughout laboratories, is the accidental omission or duplication of adding a selective supplement during sample preparation.  For clearer selective enrichment of food samples, the new Thermo Scientific Oxoid PrecisBlue Supplement has been specifically designed to reduce workflow errors. The addition of an inert blue dye provides a clear visual reference, enabling easy identification of supplemented samples.
1. What is PCR testing in food microbiology? 
PCR testing in food microbiology is a molecular method that detects pathogen DNA in food and environmental samples. It offers high sensitivity, specificity, and faster results than traditional culture methods. 
2. What is the Thermo Scientific SureTect PCR System? 
The SureTect PCR System is a rapid, validated solution for detecting foodborne pathogens. It combines ready-to-use assays with simplified workflows and fast, reliable results in under 24 hours. 
3. Which pathogens can the SureTect system detect? 
The SureTect system detects Salmonella spp., Listeria monocytogenes, Listeria spp., STEC, E. coli O157:H7, Campylobacter spp., Cronobacter spp., and Vibrio spp. 
4. What food types can be tested using SureTect PCR assays? 
Validated matrices include raw meat, poultry, seafood, dairy, infant formula, RTE foods, produce, and environmental samples like swabs and rinse water. 
5. How fast are the SureTect PCR results? 
The SureTect system delivers results in less than 24 hours, enabling faster product release and reduced inventory hold times. 
6. Is the SureTect system validated and regulatory-compliant? 
Yes, SureTect assays are AOAC-PTM, MicroVal, and AFNOR validated, compliant with ISO 16140, and support GFSI, HACCP, FSMA, and BRCGS compliance. 
7. Can SureTect workflows be automated? 
Yes, full or partial automation is supported for higher throughput, reduced manual handling, and faster lab workflows. 
8. How can I start using SureTect in my lab? 
Contact our team to request a demo or consultation. We offer onboarding, validation support, and technical assistance to get your lab running quickly. 
For Research Use Only. Not for use in diagnostic procedures.