ARL EQUINOX Range
From classroom teaching, to routine measurements, to discovery of the next breakthrough material, Thermo Scientific ARL EQUINOX X-ray Diffractometers are designed to meet the needs of today's laboratory.
- Fast acquisition with friendly interpretation data intuitive software for providing fast answers
- Quickly save your data with multiple data-export options
- Easily change samples with a large, accessible sample compartment
ARL EQUINOX XRD models are widely used in both research and industry working on polycrystalline materials: geology and minerals, materials science, energy materials, ceramics, metals and other applied materials, polymers, catalysts, chemicals, nanomaterials, pharmaceuticals, semiconductors, mining and cement, teaching, etc.
ARL EQUINOX 100 | ARL EQUINOX 1000 | |
---|---|---|
Product image | ![]() | ![]() |
Standalone benchtop XRD for routine and research labs | Benchtop XRD for routine and research labs | |
Reflection mode | ✓✓✓ | ✓✓ |
Model according to the customer needs | ||
Transmission mode | ✓✓✓ | ✓✓ |
Perfect for a benchtop model (by capillary) | ||
Phase id & QPA | ✓✓✓ | ✓✓✓ |
QA / QC CFR21 | ✓✓✓ | ✓✓✓ |
Teaching | ✓✓✓ | ✓✓ |
High & low temperature | ✓✓ * | ✓✓ * |
* Anton Paar chambers -10°C to 150°C (BTS 150) or RT to 500°C (BTS 500) | ||
Thin film GIXRD | ✓✓ | ✓✓ |
Perfect for a benchtop model | ||
Thin film XRR | According to the sample | ✓ |
High resolution | --- | --- |
Low angle | ~1° 2θ | ~1° 2θ |
ARL EQUINOX 100, ideal XRD benchtop for fast acquisition and dynamic studies
Resources
Mining|Metals
Mining|Metals
Mining|Metals