Search Thermo Fisher Scientific
This map and the annotated sequence below portrays the circular negative control vector provided with the kit. It includes the negative control siRNA template (55 base pairs) between the BamH I and Hind III sites.
Site | Enzyme |
---|---|
405 | BntI |
2528 | BsaI |
1121 | BseRI |
3447 | BsmI |
4363 | BspMI |
3393 | BstZ17I |
5198 | EcoO109I |
462 | HindIII |
235 | KasI |
1114 | KpnI |
3580 | NaeI |
236 | NarI |
3578 | NgoMIV |
401 | NheI |
406 | NotI |
1574 | PciI |
4653 | PshAI |
3506 | PsiI |
4347 | PstI |
4269 | RsrII |
3902 | SacII |
3384 | SalI |
1458 | SapI |
4972 | SexAI |
237 | SfoI |
4318 | SgfI |
4721 | SmaI |
763 | SnaBI |
4745 | StuI |
4719 | XmaI |
3066 | XmnI |
Enzyme |
---|
BsaBI |
BsgI |
BsiWI |
BsrGI |
BssHII |
BstBI |
BstEII |
BstXI |
Bsu36I |
BtrI |
ClaI |
EcoICRI |
EcoNI |
EcoRV |
FalI |
FalI' |
FseI |
FspAI |
HpaI |
MfeI |
MluI |
MscI |
NruI |
PacI |
PflMI |
PmeI |
PpuMI |
PspOMI |
PsrI |
PsrI' |
SacI |
SanDI |
SbfI |
SfiI |
SgrAI |
SpeI |
SrfI |
SwaI |
TaqII' |
XbaI |
XcmI |
XhoI |