pSilencer™ 4.1-CMV neo
pSilencer 4.1-CMV neo Vector Map
pSilencer 4.1-CMV neo Annotated Promoter/Cloning Site Region
We recommend the following primers for sequencing:
Forward: 5'-AGGCGATTAAGTTGGGTA-3'
Reverse: 5'-CGGTAGGCGTGTACGGTG-3'
pSilencer 4.1-CMV neo Restriction Sites
Enzymes with single Restriction Sites
Site | Enzyme |
---|---|
1110 | Acc65I |
1574 | AflIII |
2467 | AhdI |
539 | AloI |
507 | AloI` |
1990 | AlwNI |
4431 | AvaI |
4452 | AvrII |
515 | BanHI |
3885 | BanII |
2972 | BcgI |
3006 | BcgI` |
4401 | BclI |
1104 | BglII |
405 | BmtI |
4387 | BsaBI |
2528 | BsaI |
1449 | BsaXI |
1419 | BsaXI` |
45 | BsmBI |
3326 | BsmI |
3845 | BssHII |
4933 | EcoO109I |
1118 | EcoRI |
462 | HindIII |
1114 | KpnI |
4164 | MscI |
401 | NheI |
406 | NotI |
3562 | NruI |
1574 | PciI |
3385 | PsiI |
4197 | PstI |
3727 | RsrII |
2947 | ScaI |
4684 | SexAI |
4433 | SmaI |
763 | SnaBI |
4457 | StuI |
4127 | Tth111I |
4431 | XmaI |
3066 | XmnI |
Non-Cutting Enzymes:
Enzyme |
---|
AarI |
AccI |
AfeI |
AflII |
AgeI |
AleI |
ApaI |
AscI |
AsiSI |
BaeI |
BaeI' |
BbsI |
BbvCI |
BlpI |
BmgBI |
Bpu10I |
BsgI |
BsiWI |
BspEI |
BsrGI |
BstBI |
BstEII |
BstXI |
BstZ17I |
Bsu36I |
BtrI |
ClaI |
DraIII |
EcoICRI |
EcoNI |
EcoRV |
FseI |
FspAI |
HincII |
HpaI |
MfeI |
MluI |
PacI |
PflMI |
PmeI |
PmlI |
PpuMI |
PshAI |
PspOMI |
PsrI |
PsrI' |
SacI |
SacII |
SalI |
SanDI |
SbfI |
SfiI |
SgfI |
SgrAI |
SpeI |
SrfI |
SwaI |
TaqII' |
XbaI |
XcmI |
XhoI |
For Research Use Only. Not for use in diagnostic procedures.