Shuttle Vector 1.0-CMV Map
Shuttle Vector 1.0-CMV Map
Shuttle Vector 1.0-CMV Annotated Promoter/Cloning Site Region
We recommend the following primers for sequencing:
5' Sequencing Primer: 5'-ggatttccaagtctccac-3'
3' Sequencing Primer: 5'-agcaccttccagatctgc-3'
Detail of Multi-Cloning Site
Shuttle Vector 1.0-CMV Restriction Sites (Vector Size: 5891 bp)
Enzymes with single Restriction Sites
Site Enzyme
2978AarI
375Acc65I
1321AflII
158AflIII
3310BbeI
15BmtI
1853BsaBI
389BsaXI
359BsaXI`
5742BsrBI
207BsrGI
2974BstEII
2398BstXI
3554BstZ171
5696EarI
5123FspI
3306KasI
379KpnI
1712MfeI
3307NarI
620NdeI
11NheI
1085NotI
3899NruI
7NruI
158PciI
1959PflMI
5271PvuI
1092SalI
5381ScaI
3308SfoI
202SgrAI
726SnaBI
1029SpeI
3569StuI
1549Tth111I
970XhoI
5500XmnI
Non-Cutting Enzymes:
Enzyme
AgeI
AleI
AloI
AloI'
AscI
AsiSI
AvrII
BaeI
BaeI'
BclI
BlpI
BsiWI
BspEI
BstBI
Bsu36I
ClaI
EcoNI
EcoRI
EcoRV
FalI
FalI'
FseI
FspAI
HindIII
HpaI
MluI
PmeI
PmlI
PsiI
PsrI
PsrI'
RsrII
SanDI
SapI
SbfI
SfiI
SgfI
SrfI
SwaI
TaqII'
XbaI