forma-ref-freezer-family-230x195
Automatic defrost -30°C laboratory freezers accommodate a variety of capacities, demanding, space challenged environments, and samples and applications ranging from routine to complex.
  • Environmentally-friendly, non CFC and HCFC refrigerants
  • Energy Star certified laboratory freezers
  • Alarms built in, plus connectivity options for state-of-the-art monitoring and reporting
  • Quiet operation
  • Keyed security, including power switch and setpoint
  • Easy mobility plus lockable casters 
  • Ergonical handles
  • GMP Clean Room Class A / ISO 6 (ISO EN 14644-1) compatible 
Browse our complete portfolio or contact one of our experts to help you make a selection.
-30 high performance auto defrost lab freezers
Our auto defrost, high performance freezers are designed for applications requiring storage of reagents, pharmaceuticals, biolocal and other commonly used laboratory materials such as primers, siRNA molecules and fetal bovine serum (FBS).
Forced Air circulation for uniform temperature throughout the cabinet and fast door opening recovery, and non-invasive defrost with time and temperature guided defrost cycle eliminate the need to remove samples to manually defrost the unit.  
Looking for freezers specifically designed for plasma storage? Visit our Plasma Freezers page.  
Compact -30 auto defrost freezers
Forma Undercounter Freezer
 Designed for medical and scientific applications and featuring large, easy-to-read icon-based contols and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.
Request a quote
Capacity4.9 cu. ft. (139L)
Temperature Range−30°C
DoorsSingle, Solid
Interior (DxWxH)20.5 x 20 x 20 in. (50.8 x 50.8 x 50.8cm)
Exterior (DxWxH)33.4 x 24 x 33.4 in. (85 x 61 x 85cm)
Shipping Weight220 lb. (100kg)
NEMA 5-15P
UK
UK
UK
Revco Undercounter Freezer
 Designed for medical and scientific applications and featuring large, easy-to-read icon-based contols and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.
Request a quote
Capacity4.9 cu. ft. (139L)
Temperature Range−30°C
DoorsSingle, Solid
Interior (DxWxH)20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8cm)
Exterior (DxWxH)28.9 x 24 x 34 in. (73.4 x 61 x 86.4cm)
Shipping Weight212 lb. (96kg)
NEMA 5-15P
NEMA 5-15P
NEMA 6-15P
NEMA 6-15P
Single door -30 auto defrost freezers
Designed with V-drive technology that supports critical sample protection and sustainability objectives, and provides whisper quiet operation for clinical and laboratory-grade storage.
11.5 cu. ft. (326L) HP Lab Freezers
23 cu. ft. (650L) HP Lab Freezers
29.2 cu. ft. (827L) HP Lab Freezers
Request a quote
Capacity11.5 cu. ft. (326L)23 cu. ft. (650L)29.2 cu. ft. (827L)
Temperature Range−35° to −15°C−35° to −15°C−35° to −15°C
DoorsSingle, solidSingle, solidSingle, solid
Interior (DxWxH)21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1cm)28.5 x 24 x 58 in. (72.3 x 61 x 147.3cm)28.5 x 30.0 x 58.0 in. (72.3 x 76.2 x 147.3cm)
Exterior (DxWxH)31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4cm)37.8 x 28 x 78.6 in. (96 x 71.1 x 199.6cm)37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4cm)
Shipping Weight335 lb. (152kg)423 lb. (198 kg)439 lb. (199kg)
Designed for medical and scientific applications and featuring large, easy-to-read icon-based contols and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.
11.5 cu. ft. (326L) HP Lab Freezers
29.2 cu. ft. (827L) HP Lab Freezers
Request a quote
Capacity11.5 cu. ft. (326L)29.2 cu. ft. (827L)
Temperature Range−30°C−30°C
DoorsSingle, solidSingle, solid
Interior (DxWxH)21.75 x 20 x 52.4 in. (50.8 x 50.8 x 133.1cm)29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Exterior (DxWxH)29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm)37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)
Shipping Weight280 lb. (100kg)445 lb. (202kg)
Single, solid
Designed for medical and scientific applications and featuring large, easy-to-read icon-based contols and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.
11.5 cu. ft. (326L) HP Lab Freezers
Request a quote
Capacity11.5 cu. ft. (326L)
Temperature Range−30°C
DoorsSingle, Solid
Interior (DxWxH)21 x 20 x 52.4 in. (53.3 x 50.8 x 133.1cm)
Exterior (DxWxH)29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm)
Shipping Weight280 lb. (127kg)
Designed for medical and scientific applications and featuring large, easy-to-read icon-based controls and displays, microprocessor control and scratch-resistant cold-rolled steel interior and exterior finishes.
11.5 cu. ft. (326L) HP Lab Freezers
29.2 cu. ft. (827L) HP Lab Freezers
Request a quote
Capacity11.5 cu. ft. (326L)29.2 cu. ft. (827L)
Temperature Range−30°C−30°C
DoorsSingle, SolidSingle, Solid
Interior (DxWxH)20 x 21.75 x 52.4 in. (50.8 x 53.3 x 133.1cm)30 x 29 x 58 in. (76.2 x 73.7 x 147.3cm)
Exterior (DxWxH)29.3 x 30 x 73.6 in. (74.3 x 76.2 x 186.9cm)34 x 37.2 x 79.2 in. (86.4 x 94.5 x 201.2cm)
Shipping Weight280 lb. (127kg)445 lb. (202kg)
Double door -30 auto defrost freezers
Designed with features that support sample protection and sustainability objectives with significant energy savings and whisper quiet operation for clinical and laboratory-grade storage.
TSX Series HP Lab Freezers
51.1 cu. ft. (1447L)
Request a quote
Capacity51.1 cu. ft. (1447L)
Temperature Range-35° to -15°C
DoorsDouble, Solid
Interior (DxWxH)28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3cm)
Exterior (DxWxH)37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4cm)
Shipping Weight621 lb. (282L)
-20 auto defrost lab freezers
General-purpose lab storage for routine samples with capacities ranging from 265L to 1,048L.
Not available in North America or other 60Hz regions.
400L GPS Series Single Door Lab Freezers
700L GPS Series Single Door Lab Freezers
1400L GPS Series Double Door Lab Freezers
Request a quote
Capacity400L700L1400L
Temperature Range−12° to −25°C−12° to −25°C−12° to −25°C
DoorsSingle, SolidSingle, SolidDouble, Solid
Interior (DxWxH)19.7 x 18.9 x 45.3 in. (50 x 48 x 115cm)26 x 26 x 51.1 in. (66 x 66 x 130cm)26 x 51.1 x 51.1 in. (66 x 130 x 130cm)
Exterior (DxWxH)23.6 x 24.8 x 74.8 in. (60 x 63 x 190cm)31.9 x 27.6 x 78.0 in. (81 x 70 x 198cm)31.5 x 55.1 x 78.0 in. (80 x 140 x 198cm)
Shipping Weight95kg130kg200kg
Single, Solid
Single, Solid
Which plug type do I need for my auto defrost freezer?
 Standard Plug TypeVoltageHz
NEMA 5-15115–12060
NEMA 5-20115–12060
NEMA 6-15208–23060
Continental EU
23050
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic