Designed with sample protection as a priority
Your laboratory oven is an invaluable part of your daily workflow, enabling simple glassware drying to the most complex, controlled heating applications. Our heating and drying ovens portfolio ensures temperature stability and reproducibility for all your desired application needs. Helping to enable maximum efficiency, safety, and ease of use, Thermo Scientific Heratherm heating and drying ovens are designed with sample protection as a top priority.
Browse our complete portfolio or let one of our experts help you make a selection.
Understanding Gravity and Mechanical convection
How Gravity Convection works
In a gravity convection oven, the temperature distribution is based on warm air moving upwards (see above). There is no fan that actively distributes the air inside the chamber. The benefit of this technology is very low air turbulences for gentle drying and heating.
How Mechanical Convection works
In a mechanical convection (or forced air) oven an integrated fan actively moves the air inside the chamber – resulting in an even temperature distribution throughout the chamber (see above). One benefit is optimal temperature uniformity for reproducible results, for example in material testing and for drying protocols with very tight temperature requirements. Another advantage is a much faster drying process compared to gravity convection. When opening the door, the temperature will recover faster to the set temperature level in a mechanical convection oven.
Top reasons to invest in our lab heating and drying ovens
- Energy efficient, increased cost effectiveness, reduced environmental impact
- Unique door design reduces heat emission
- Timer for effective usage and additional energy savings
- Space saving footprint without compromising volume
- Flexible shelving system allows for optimal space utilization
- Smart safety system surveys functionality of oven consistently
- Temperature uniformity as good as +/-1.3°C at 150°C
- Automatic audible and visual overtemperature alarm
- Access port for data logging with independent sensors
- Low external surface temperature for easy handling
- Easy-to-read, large vacuum fluorescent display
- Easy setting with intuitive interface
- Easy cleaning with rounded corners
|
|
See chart below for unit specific features
General Protocol | Advanced Protocol | Advanced | |||
![]() | ![]() | ![]() | |||
Size | Table Top | Large Capacity | Table Top | Large Capacity | Table Top |
Temperature range | 50°C - 250℃ | 50°C - 330°C | 50°C - 250℃ | 50°C - 330°C | |
Temperature uniformity/stability | Good Performance | Best Performance | Best Performance | ||
Gravity convection | choice | ✓ | choice | • | choice |
Mechanical convection | choice | • | choice | ✓ | choice |
Adjustable fan speed | • | 5 speeds | 2 speeds | 5 speeds | |
Timer function | On / Off | Choice of weekly, hourly or real time | Choice of weekly, hourly or real time | ||
Programmable (ramps/dwells) | • | ✓ | ✓ | ||
Boost function for accelerated heat-up | • | ✓ | • | ✓ | |
Automatic undertemperature alarm | • | • | ✓ | ||
Door alarm | • | • | ✓ | ✓ | |
Lockable door | • | • | ✓ | ||
Exterior Finish | Coated Steel | Coated Steel | Coated Steel or Stainless Steel | ||
Products by electrical requirements |
Stainless Steel or |
Stainless Steel or |
Lab heating and drying ovens for special applications
Specialty Oven | Application |
Precision High-Performance Ovens | Benefit from the uniform heating, precise temperature control and fast drying of these high-performance ovens. |
Precision Compact Ovens | Conserve valuable benchtop space with a compact heating and drying oven featuring mechanical or gravity convection. Ideal for drying and baking applications. |
Flammable solvent |
Flammable solvent |
Heratherm ovens are available in mechanical models with superior temperature control and uniformity; or with gentle gravity convection, perfect for materials sensitive to airflow.
Learn more about Heratherm ovens.
Resources
Support
- Unity Lab Services
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services