Search Thermo Fisher Scientific
A sterile, liquid medium prepared with 200 µM calcium chloride for the culture of human epidermal keratinocytes. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use. Below is the composition list. The concentrations are proprietary.
Catalog Number(s)
M154500Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | confidential | n/a |
L-Alanine | 89.0 | confidential | n/a |
L-Arginine hydrochloride | 211.0 | confidential | n/a |
L-Asparagine-H2O | 150.0 | confidential | n/a |
L-Aspartic acid | 133.0 | confidential | n/a |
L-Cysteine | 121.0 | confidential | n/a |
L-Glutamic Acid | 147.0 | confidential | n/a |
L-Glutamine | 146.0 | confidential | n/a |
L-Histidine hydrochloride-H2O | 210.0 | confidential | n/a |
L-Isoleucine | 131.0 | confidential | n/a |
L-Leucine | 131.0 | confidential | n/a |
L-Lysine hydrochloride | 183.0 | confidential | n/a |
L-Methionine | 149.0 | confidential | n/a |
L-Phenylalanine | 165.0 | confidential | n/a |
L-Proline | 115.0 | confidential | n/a |
L-Serine | 105.0 | confidential | n/a |
L-Threonine | 119.0 | confidential | n/a |
L-Tryptophan | 204.0 | confidential | n/a |
L-Tyrosine | 181.0 | confidential | n/a |
L-Valine | 117.0 | confidential | n/a |
Vitamins | |||
Choline chloride | 140.0 | confidential | n/a |
D-Pantothenic Acid | 238.0 | confidential | n/a |
Folic Acid | 441.0 | confidential | n/a |
Myo-Inositol | 180.0 | confidential | n/a |
Niacinamide | 122.0 | confidential | n/a |
Pyridoxal hydrochloride | 206.0 | confidential | n/a |
Riboflavin | 376.0 | confidential | n/a |
Thiamine hydrochloride | 337.0 | confidential | n/a |
Vitamin B12 | 1355.0 | confidential | n/a |
d-Biotin | 244.0 | confidential | n/a |
Inorganic Salts | |||
Ammonium Metavanadate NaVO3 | 117.0 | confidential | n/a |
Ammonium Molybdate ((NH4)6Mo7O24-4H2O) | 1236.0 | confidential | n/a |
Calcium Chloride (CaCl2-2H2O) | 147.0 | confidential | n/a |
Cupric sulfate (CuSO4-5H2O) | 250.0 | confidential | n/a |
Ferric sulfate (FeSO4-7H2O) | 278.0 | confidential | n/a |
Magnesium Chloride (MgCl2-6H2O) | 203.0 | confidential | n/a |
Manganese Sulfate (MnSO4-H20) | 169.0 | confidential | n/a |
Nickelous Chloride NiCl2 6H2O | 238.0 | confidential | n/a |
Potassium Chloride (KCl) | 75.0 | confidential | n/a |
Sodium Acetate (NaCH3COO.3H2O) | 136.0 | confidential | n/a |
Sodium Bicarbonate (NaHCO3) | 84.0 | confidential | n/a |
Sodium Chloride (NaCl) | 58.0 | confidential | n/a |
Sodium Meta Silicate Na2SiO3 9H2O | 284.0 | confidential | n/a |
Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | confidential | n/a |
Sodium Selenite (Na2SeO3) | 173.0 | confidential | n/a |
Tin Chloride (SnCl2.2H2O) | 226.0 | confidential | n/a |
Zinc sulfate (ZnSO4-7H2O) | 288.0 | confidential | n/a |
Other Components | |||
Adenine.HCl | 172.0 | confidential | n/a |
D-Glucose (Dextrose) | 180.0 | confidential | n/a |
DL-alpha-Lipoic Acid | 206.0 | confidential | n/a |
Ethanolamine | 61.0 | confidential | n/a |
HEPES | 238.0 | confidential | n/a |
O-Phosphorylethanolamine | 141.0 | confidential | n/a |
Phenol Red | 376.4 | confidential | n/a |
Putrescine 2HCl | 161.0 | confidential | n/a |
Sodium Pyruvate | 110.0 | confidential | n/a |
Thymidine | 242.0 | confidential | n/a |
Medium 106
Cat No:
M106500
Medium 131
Cat No:
M131500
Medium 154
Cat No:
M154CF500
Medium 154
Cat No:
M154CFPRF500
EpiLife® Medium
Cat No:
MEPI500CA
EpiLife® Medium
Cat No:
MEPICF500
EpiLife® Medium
Cat No:
MEPICFPRF500
Medium 231
Cat No:
M231500
Medium 200
Cat No:
m200500
Medium 200
Cat No:
M200PRF500
Medium 254
Cat No:
M254500
Medium 254
Cat No:
M254CF500
Medium 171
Cat No:
M171500