Technical Resources - Media Formulations
12492 - Advanced MEM
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.07.50.1
L-Alanine89.08.90.099999994
L-Arginine hydrochloride211.0126.00.5971564
L-Asparagine132.013.20.1
L-Aspartic acid133.013.30.1
L-Cystine 2HCl313.031.00.09904154
L-Glutamic Acid147.014.70.1
L-Histidine hydrochloride-H2O210.042.00.2
L-Isoleucine131.052.00.39694658
L-Leucine131.052.00.39694658
L-Lysine hydrochloride183.072.50.39617488
L-Methionine149.015.00.10067114
L-Phenylalanine165.032.00.19393939
L-Proline115.011.50.1
L-Serine105.010.50.1
L-Threonine119.048.00.40336135
L-Tryptophan204.010.00.04901961
L-Tyrosine disodium salt dihydrate261.052.00.19923371
L-Valine117.046.00.3931624
Vitamins
Ascorbic Acid phosphate290.02.50.00862069
Choline chloride140.01.00.007142857
D-Calcium pantothenate477.01.00.002096436
Folic Acid441.01.00.0022675737
Niacinamide122.01.00.008196721
Pyridoxine hydrochloride204.01.00.004901961
Riboflavin376.00.12.6595744E-4
Thiamine hydrochloride337.01.00.002967359
i-Inositol180.02.00.011111111
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0200.01.8018018
Magnesium Sulfate (MgSO4) (anhyd.)120.097.670.8139166
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Bicarbonate (NaHCO3)84.02200.026.190475
Sodium Chloride (NaCl)58.06800.0117.24138
Sodium Phosphate dibasic (Na2HPO4-H2O)138.0140.01.0144928
Proteins
AlbuMAX® II400.0Infinity
Human Transferrin (Holo)7.5Infinity
Insulin Recombinant Full Chain5964.010.00.0016767271
Trace Elements
Ammonium Metavanadate117.03.0E-42.5641027E-6
Cupric Sulfate250.00.001255.0E-6
Manganous Chloride198.05.00.025252525
Sodium Selenite173.00.0052.8901733E-5
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Ethanolamine98.01.90.019387756
Glutathione (reduced)307.01.00.0032573289
Phenol Red37604.010.02.6592915E-4
Sodium Pyruvate110.0110.01.0
Reference:
Related Products:
12491 - Advanced DMEM
Cat No: 12491015 , 12491023
12633 - Advanced RPMI 1640
Cat No: 12633012 , 12633020
12634 - Advanced D-MEM/F-12
Cat No: 12634010 , 12634028