Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 50.0 | 0.6666667 |
| Hydroxy L-proline | 131.0 | 10.0 | 0.07633588 |
| L-Alanine | 89.0 | 25.0 | 0.28089887 |
| L-Arginine hydrochloride | 211.0 | 70.0 | 0.33175355 |
| L-Aspartic acid | 133.0 | 30.0 | 0.22556391 |
| L-Cysteine hydrochloride-H2O | 176.0 | 260.0 | 1.4772727 |
| L-Cystine | 240.0 | 26.0 | 0.108333334 |
| L-Glutamic Acid | 147.0 | 75.0 | 0.5102041 |
| L-Histidine hydrochloride-H2O | 210.0 | 20.0 | 0.0952381 |
| L-Isoleucine | 131.0 | 20.0 | 0.15267175 |
| L-Leucine | 131.0 | 60.0 | 0.45801526 |
| L-Lysine hydrochloride | 183.0 | 70.0 | 0.38251367 |
| L-Methionine | 149.0 | 15.0 | 0.10067114 |
| L-Phenylalanine | 165.0 | 25.0 | 0.15151516 |
| L-Proline | 115.0 | 40.0 | 0.3478261 |
| L-Serine | 105.0 | 25.0 | 0.23809524 |
| L-Threonine | 119.0 | 30.0 | 0.25210086 |
| L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
| L-Tyrosine disodium salt dihydrate | 262.0 | 58.0 | 0.22137405 |
| L-Valine | 117.0 | 25.0 | 0.21367522 |
| Vitamins | |||
| Ascorbic Acid | 176.0 | 50.0 | 0.2840909 |
| Biotin | 244.0 | 0.01 | 4.0983607E-5 |
| Cholesterol | 387.0 | 0.2 | 5.1679584E-4 |
| Choline chloride | 140.0 | 0.5 | 0.0035714286 |
| D-Calcium pantothenate | 477.0 | 0.01 | 2.096436E-5 |
| Folic Acid | 441.0 | 0.01 | 2.2675737E-5 |
| Niacinamide | 122.0 | 0.025 | 2.0491803E-4 |
| Nicotinic acid (Niacin) | 123.0 | 0.025 | 2.0325204E-4 |
| Para-Aminobenzoic Acid | 137.0 | 0.05 | 3.6496352E-4 |
| Pyridoxal hydrochloride | 204.0 | 0.025 | 1.2254903E-4 |
| Pyridoxine hydrochloride | 206.0 | 0.025 | 1.21359226E-4 |
| Riboflavin | 376.0 | 0.01 | 2.6595744E-5 |
| Thiamine hydrochloride | 337.0 | 0.01 | 2.967359E-5 |
| i-Inositol | 180.0 | 0.05 | 2.7777778E-4 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.7 | 0.81416667 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 6365.0 | 109.74138 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
| Other Components | |||
| 2'Deoxyadenosine | 251.0 | 10.0 | 0.03984064 |
| 2'Deoxycytidine | 227.0 | 10.0 | 0.04405286 |
| 2'Deoxyguanosine | 267.0 | 10.0 | 0.037453182 |
| 5-Methyl-deoxycytidine | 225.0 | 0.1 | 4.4444445E-4 |
| Co-carboxylase | 461.0 | 1.0 | 0.0021691974 |
| Coenzyme A | 768.0 | 2.5 | 0.0032552083 |
| D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
| Diphosphopyridine nucleotide (NAD) | 663.0 | 7.0 | 0.01055807 |
| FAD (flavin adenine dinucleotide) | 786.0 | 1.0 | 0.0012722646 |
| Glutathione (reduced) | 307.0 | 10.0 | 0.03257329 |
| Phenol Red | 376.4 | 20.0 | 0.053134963 |
| Sodium acetate-3H2O | 136.0 | 83.0 | 0.6102941 |
| Sodium glucuronate-H2O | 236.0 | 4.2 | 0.01779661 |
| Thymidine | 242.0 | 10.0 | 0.041322313 |
| Triphosphopyridine Nucleotide (NADP) | 743.0 | 1.0 | 0.001345895 |
| Tween 80® | 5.0 | Infinity | |
| Uridine 5'- triphosphate | 484.0 | 1.0 | 0.0020661156 |
1. Parker R.C., et al. (1957) Special Publications, N.Y. Academy of Sciences, 5, 303.
21530 - CMRL, no Glutamine
Cat No:
21530027
,
21530076
21540 - CMRL, 10x, no Glutamine
Cat No:
21540026