Search
A section of mouse intestine was stained with a combination of fluorescent stains. Fibronectin, an extracellular matrix adhesion molecule, was labeled using a chicken primary antibody against fibronectin and visualized using green-fluorescent Alexa Fluor® 488 Goat Anti–Chicken IgG antibody (A11039). The filamentous actin (F-actin) prevalent in the brush border was stained with red-fluorescent Alexa Fluor® 568 phalloidin (A12380). Finally, the nuclei were stained with DAPI (D1306, D3571, D21490).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Endothelial cells. Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›