Search
Human iPSCs cultured under feeder-free conditions in StemPro® hESC Medium (A1000701) were fixed and permed using the Image-IT® Fixation/Permeabilization Kit (R37602). Cells were stained using mouse anti-Tra1-60 primary antibody, Alexa Fluor® 594 Goat Anti-Mouse (A11005) and NucBlue™ Fixed Cell Stain (R37606). Following staining, slides were mounted in ProLong® Gold Antifade reagent (P36934) and imaged on FLoid™ Cell Imaging Station (4471136).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Human iPSC staining using mouse anti beta-tubulin monoclonal antibody. Go ›
Human iPSC staining using Mouse Anti Beta-Tubulin Monoclonal Antibody. Go ›