Search
Comet assay with SYBR® Green I Nucleic Acid Gel Stain (Cat. No. S7563, S7567, S7585). DNA fragmentation associated with oxidative DNA damage was visualized using Trevigen's CometAssay kit. HL-60 cells were treated with hydrogen peroxide and immobilized onto a Trevigen CometSlide for analysis. The cells were gently lysed, washed, and treated with endonuclease. Slides were subjected to electrophoresis in alkaline electrophoresis buffer and stained with SYBR® Green I stain.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›