Search
A bovine pulmonary artery endothelial cell labeled with mouse monoclonal anti–a-tubulin antibody (Cat. No. A11126) in combination with Alexa Fluor® 568 goat anti–mouse IgG (Cat. No. A11004) to stain microtubules. The image was acquired using a bandpass filter set appropriate for X-rhodamine.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›