Search
Mouse fibroblast microtubules labeled with a mouse monoclonal anti–ß-tubulin antibody in conjunction with biotin-XX goat anti–mouse IgG antibody (Cat. no. B2763) and then developed for visualization with alkaline phosphatase–mediated techniques using the ELF® 97 Cytological Labeling Kit (Cat. no. E6603). This kit's novel ELF® 97 phosphatase substrate yields a yellow-green–fluorescent precipitate at the site of alkaline phosphatase activity. Prior to antibody labeling, mouse fibroblasts were fixed and permeabilized in the presence of cytoskeletal stabilizing buffer and treated with paclitaxel (Cat. no. P3456) to stabilize microtubule structures. The image was deconvolved using Huygens software (Scientific Volume Imaging, http://www.svi.nl/).
Prostate carcinoma. Go ›
Transformed mouse fibroblasts (CRE BAG 2 cells). Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›