Bovine pulmonary artery endothelial cells. Anti–bovine a-tubulin, fluorescein goat anti–mouse IgG conjugate and anti–fluorescein/Oregon Green®, R-phycoerythrin conjugate.
Microtubules of fixed bovine pulmonary artery endothelial cells (BPAEC) labeled with mouse monoclonal anti–a-tubulin antibody (Cat. no. A11126), followed by fluorescein goat anti–mouse IgG (Cat. no. F2761). The orange-fluorescent signal was produced using R-phycoerythrin–labeled anti–fluorescein/Oregon Green® antibody (Cat. no. A21250).
Microtubules of fixed bovine pulmonary artery endothelial cells (BPAEC) labeled with mouse monoclonal anti–a-tubulin antibody (Cat. no. A11126), followed by fluorescein goat anti–mouse IgG (Cat. no. F2761). The orange-fluorescent signal was produced using R-phycoerythrin–labeled anti–fluorescein/Oregon Green® antibody (Cat. no. A21250).
Related Products
Related Images