Fixed, permeabilized bovine pulmonary artery endothelial cells visualized using components of the SelectFX® Nuclear Labeling Kit and Alexa Fluor® phalloidin conjugates.
Fixed, permeabilized bovine pulmonary artery endothelial cells visualized using components of the SelectFX® Nuclear Labeling Kit (Cat. no. S33025) and Alexa Fluor® phalloidin conjugates. Nuclei and F-actin were stained, respectively, with (top left) DAPI and Alexa Fluor® 680 phalloidin (Cat. no. A22286), (top right) SYTOX® Green dye and Alexa Fluor® 568 phalloidin (Cat. no. A12380), (bottom left) 7-AAD and Alexa Fluor® 488 phalloidin (Cat. no. A12379), and (bottom right) TO-PRO-3 dye and Alexa Fluor® 350 phalloidin (Cat. no. A22281).
Fixed, permeabilized bovine pulmonary artery endothelial cells visualized using components of the SelectFX® Nuclear Labeling Kit (Cat. no. S33025) and Alexa Fluor® phalloidin conjugates. Nuclei and F-actin were stained, respectively, with (top left) DAPI and Alexa Fluor® 680 phalloidin (Cat. no. A22286), (top right) SYTOX® Green dye and Alexa Fluor® 568 phalloidin (Cat. no. A12380), (bottom left) 7-AAD and Alexa Fluor® 488 phalloidin (Cat. no. A12379), and (bottom right) TO-PRO-3 dye and Alexa Fluor® 350 phalloidin (Cat. no. A22281).
Related Products
Related Images