Search
Bovine pulmonary artery endothelial cells were labeled with fluorescein phalloidin (Cat. no. F432), which labels filamentous actin, and placed under constant illumination on the microscope with a FITC filter set using a 60× objective. Images were acquired at one-second intervals for 30 seconds. Under these illumination conditions, fluorescein photobleached to about 12% of its initial value in 30 seconds in PBS (left), but stayed at the initial value under the same illumination conditions when mounted using the reagents in the ProLong® Antifade Kit (right, Cat. no. P7481).
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›