Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Troubleshooting page
Browse our FAQ database for
more information ›
General
The QuantStudio™ 3 Real-Time PCR System is a 4-color instrument with a 3-zone VeriFlex™ block, while the QuantStudio™ 5 Real-Time PCR System is a 6-color instrument with 6-VeriFlex™ zones in a 96-well block, (or 5-color format, no VeriFlex™ with 384-well block). 
Yes, the QuantStudio™ 5 Real-Time PCR System has SAE (Security/Auditing/e-Signature) features that can be enabled in the QuantStudio™ Design and Analysis Desktop Software. 
Yes, you can purchase the QuantStudio™ 3 or QuantStudio™ 5 Real Time PCR System without a computer. In this configuration, you can run the instrument through the touchscreen or the Design and Analysis application from Thermo Fisher Cloud. 
When your new QuantStudio™ 3 or QuantStudio™ 5 Real-Time PCR System arrives in your lab, it will be factory calibrated. The recommended calibration schedule is to recalibrate every 2 years. 
The block drawer is opened and closed by using the ‘eject’ button on the touchscreen. Do not push or pull on the instrument drawer. 
Yes. To have the run file automatically upload to your Thermo Fisher Cloud account once complete, you will need to set the file destination on the touchscreen, prior to starting the run: 
  1. Before you start the run, select the ‘Properties’ tab from an open experiment 2. Touch ‘Edit’ 
  2. Data Destination - Choose USB or Cloud 
Alternatively, you could use Cloud Connect Utility to have all files from a specific hard drive folder location automatically upload to the cloud (for use with the QuantStudio™ Design and Analysis Desktop Software). 
Yes, you can do this from either the instrument touchscreen, or from the Design and Analysis application of the Applied Biosystems™ qPCR Analysis Modules. You can also monitor the plots in real time with the QuantStudio™ Design and Analysis Desktop Software v1.2 or later.
We recommend using our MeltDoctor HRM Calibration plates. Use the plate type that matches the block on your QuantStudio 3 or QuantStudio 5 Real-Time PCR System:
0.1 mL, 96-Well: Cat. No. 4425618
384-Well: Cat. No. 4425559
0.2 mL, 96-well: Cat. Nos. 4425562 and 4425557 (Note: We do not have a 0.2 mL calibration plate, but you can make one by combining the MeltDoctor HRM Calibration Standard with the MeltDoctor HRM Reagent Kit)
To set up the QuantStudio 3 or QuantStudio 5 Real-Time PCR System for WiFi capability, we recommend following the instructions in the manual (https://tools.thermofisher.com/content/sfs/manuals/MAN0010407_QuantStudio3_5_InstallUseMaint_UG.pdf). You will also need to purchase the High Power USB WiFi Module (Cat. No. A26774).
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Optical Caps on the ProFlex, SimpliAmp, and Veriti thermal cycler 96-well blocks, and QuantStudio 3 and QuantStudio5 blocks.
The MicroAmp Optical 8-Tube Strip with Attached Optical Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well PCR System
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
- 7000 PCR System
- 7300 Real-Time PCR System
- 7500 Real-Time PCR system
- QuantStudio Real-Time PCR Systems
- ViiA7Real-Time PCR System, 96-well
The MicroAmp 8-Tube Strip with Attached Domed Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well 
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Domed Caps on the ProFlex, SimpliAmp, and Veriti thermal cycler 96-well blocks.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
These new strips have new features including a graduated 20 μL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
These new strips have new features including a graduated 20 μL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Please refer to the table on Page 2 of this flyer for compatibility information.
Please refer to the table on Page 2 of this flyer for compatibility information.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
These new strips have new features including a graduated 20 μL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
These new strips have new features including a graduated 20 μL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Operation from Thermo Fisher Cloud
You can set up your run in the Design and Analysis application, but to actually start the run, you will have to go to the instrument touchscreen. 
  1.  Set up the run in the cloud from the Design and Analysis application within the Applied Biosystems™ qPCR Analysis Modules. 
  2. Save the experiment before starting a run: The file name was created from the experiment Name entered in the Properties tab. However, changes to the experiment Name in the Properties tab after the first save do not update the file name. To change the file name before a run, select Actions --> Save As.
  3.  Go to an available cloud-connected instrument, enter your PIN to access this file, and start the run by choosing ‘Open Template’. Following a run, view experiment details, run duration, and instrument and software information in the Run Control screen of the Run tab. 
Data Analysis
Use the Applied Biosystems™ qPCR Analysis Module (RQ application) with your Thermo Fisher Cloud account. 
Yes, you can perform HRM experiments on the QuantStudio 3 and QuantStudio 5 Real-Time PCR Systems. The data can either be analyzed in the HRM v3.1 Software (Cat. No. A30150), or in the Thermo Fisher Cloud HRM Analysis Application. Please see here (https://tools.thermofisher.com/content/sfs/manuals/MAN0014394_HighResMeltExperiment_GSG.pdf) for the user manual (with Cloud workflow). Note: QuantStudio 3/QuantStudio 5 Real-Time PCR System firmware version 1.1 or later is required to perform HRM experiments.
Need more information? Contact us ›