Learn how the Vanquish Neo UHPLC system can bring value to your lab.
Discover why the Vanquish Neo UHPLC system is the new standard in low-flow LC.
Learn how the Vanquish Neo UHPLC system accelerates new discoveries and productivity.
Learn how tandem direct injection can help you get more from your Vanquish Neo UHPLC system by increasing sample capacity without compromising performance.
All-in-one low-flow UHPLC system
  • ProFlow XR technology delivers excellent retention time precision from nano up to 100 μL/min at pressures up to 1500 bar
  • Industry-leading flow-pressure footprint enables high-resolution separations on ultra-long columns and high throughput
 Learn more
Maximum performance
  • Next-generation SmartInject technology drives retention time precision, column lifetime, and ESI stability
  • Automated multi-wash procedures improve low-level analyte sensitivity
  • Vial bottom detection enables precise and accurate injections with minimal sample waste
 Learn more
Enhanced productivity
  • Fast sample loading and column equilibration improves sample throughput and MS utilization
  • Unique split-loop autosampler design minimizes sample dispersion and sample waste while enhancing workflow versatility
  • The tandem direct injection workflow provides maximized sample throughput and mass spectrometer utilization
 Learn more
Versatile LC-MS workflows
  • Seamless integration with the Thermo Scientific mass spectrometry portfolio through a comprehensive suite of ESI sources
  • Optimized configurations including direct and tandem direct injection, and trap-and-elute workflows ensure superior performance and system versatility
 Learn more
Intelligent operation
  • The Vanquish User Interface improves your user experience by providing walk-up and remote system monitoring, control, and automated procedures
  • The method editor wizard ensures compatibility of method parameters with installed consumables
 Learn more
All-in-one low-flow UHPLC system
  • ProFlow XR technology delivers excellent retention time precision from nano up to 100 μL/min at pressures up to 1500 bar
  • Industry-leading flow-pressure footprint enables high-resolution separations on ultra-long columns and high throughput
 Learn more
Maximum performance
  • Next-generation SmartInject technology drives retention time precision, column lifetime, and ESI stability
  • Automated multi-wash procedures improve low-level analyte sensitivity
  • Vial bottom detection enables precise and accurate injections with minimal sample waste
 Learn more
Enhanced productivity
  • Fast sample loading and column equilibration improves sample throughput and MS utilization
  • Unique split-loop autosampler design minimizes sample dispersion and sample waste while enhancing workflow versatility
  • The tandem direct injection workflow provides maximized sample throughput and mass spectrometer utilization
 Learn more
Versatile LC-MS workflows
  • Seamless integration with the Thermo Scientific mass spectrometry portfolio through a comprehensive suite of ESI sources
  • Optimized configurations including direct and tandem direct injection, and trap-and-elute workflows ensure superior performance and system versatility
 Learn more
Intelligent operation
  • The Vanquish User Interface improves your user experience by providing walk-up and remote system monitoring, control, and automated procedures
  • The method editor wizard ensures compatibility of method parameters with installed consumables
 Learn more
Drive your science forward by incorporating the Vanquish Neo UHPLC system into a workflow with other Thermo Scientific products, including our low-flow (U)HPLC columns, industry-leading mass spectrometers, and a software platform that streamlines your laboratory processes.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes