Synchronized synergy
Thermo Scientific Orbitrap Astral Mass Spectrometers are powered by unique architecture featuring a quadrupole mass analyzer and two high-resolution accurate mass (HRAM) analyzers: the Thermo Scientific Orbitrap analyzer and Thermo Scientific Astral analyzer. This powerful combination enables novel data acquisition strategies, including narrow-window data-independent acquisition (nDIA) and simultaneous quantitation and discovery (SQUAD) workflows, to deliver higher performance across many applications.
Accelerate discoveries in proteomics, metabolomics, lipidomics, structural biology, and biopharma applications with faster throughput, deeper coverage, higher sensitivity, and accurate and precise quantitation to achieve the next generation of breakthroughs.
The Orbitrap Astral Mass Spectrometer family
Thermo Scientific Orbitrap Astral Zoom Mass Spectrometer
Thermo Scientific Orbitrap Astral Mass Spectrometer
Our General Purpose product-lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks.
For General Lab Use Only - Not For Diagnostic Procedures.