Product References
Histological changes of cervical tumours following Zanthoxylum acanthopodium DC treatment, and its impact on cytokine expression.
Saudi journal of biological sciences
Simanullang RH,Situmorang PC,Herlina M,Silalahi B,Manurung SS
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Immunohistochemistry
Fri Apr 01 00:00:00 EDT 2022
Circulating Regulatory B-Lymphocytes in Patients with Acute Myocardial Infarction: A Pilot Study.
Journal of cardiovascular development and disease
Volodarsky I,Shimoni S,Haberman D,Mirkin V,Fabrikant Y,Yoskovich Mashriki T,Zalik A,George J
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Thu Dec 22 00:00:00 EST 2022
Induction of cytotoxic effector cells towards cholangiocellular, pancreatic, and colorectal tumor cells by activation of the immune checkpoint CD40/CD40L on dendritic cells.
Cancer immunology, immunotherapy : CII
Sadeghlar F,Vogt A,Mohr RU,Mahn R,van Beekum K,Kornek M,Weismüller TJ,Branchi V,Matthaei H,Toma M,Schmidt-Wolf IGH,Kalff JC,Strassburg CP,González-Carmona MA
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in ELISA
Sat May 01 00:00:00 EDT 2021
Early loss of T lymphocyte 4-1BB receptor expression is associated with higher short-term mortality in alcoholic hepatitis.
PloS one
Eriksen LL,Nielsen MA,Laursen TL,Deleuran B,Vilstrup H,Støy S
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Mon Nov 29 00:00:00 EST 2021
Cysticercus cellulosae Regulates T-Cell Responses and Interacts With the Host Immune System by Excreting and Secreting Antigens.
Frontiers in cellular and infection microbiology
Fan X,Zhang Y,Ouyang R,Luo B,Li L,He W,Liu M,Jiang N,Yang F,Wang L,Zhou B
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Wed Oct 20 00:00:00 EDT 2021
Altered T-Cell Subsets are Associated with Dysregulated Cytokine Secretion of CD4+ T Cells During HIV Infection.
Journal of inflammation research
Wang D,Jiang Y,Song Y,Zeng Y,Li C,Wang X,Liu Y,Xiao J,Kong Y,Zhao H
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Wed Apr 27 00:00:00 EDT 2022
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
Multiple Sclerosis-Associated Changes in the Composition and Immune Functions of Spore-Forming Bacteria.
mSystems
Cekanaviciute E,Pröbstel AK,Thomann A,Runia TF,Casaccia P,Katz Sand I,Crabtree E,Singh S,Morrissey J,Barba P,Gomez R,Knight R,Mazmanian S,Graves J,Cree BAC,Zamvil SS,Baranzini SE
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
β-Catenin Stabilization in Skin Fibroblasts Causes Fibrotic Lesions by Preventing Adipocyte Differentiation of the Reticular Dermis.
The Journal of investigative dermatology
Mastrogiannaki M,Lichtenberger BM,Reimer A,Collins CA,Driskell RR,Watt FM
53-7108 was used in Flow cytometry/Cell sorting to study how Wnt/β-catenin signalling may control adipogenic cell fate within the lower dermis, which potentially contributes to the pathogenesis of fibrotic skin diseases.
Wed Jun 01 00:00:00 EDT 2016
Functional expression of aryl hydrocarbon receptor on mast cells populating human endometriotic tissues.
Laboratory investigation; a journal of technical methods and pathology
Mariuzzi L,Domenis R,Orsaria M,Marzinotto S,Londero AP,Bulfoni M,Candotti V,Zanello A,Ballico M,Mimmi MC,Calcagno A,Marchesoni D,Di Loreto C,Beltrami AP,Cesselli D,Gri G
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Immunofluorescence
Thu Sep 01 00:00:00 EDT 2016
Functional expression of aryl hydrocarbon receptor on mast cells populating human endometriotic tissues.
Laboratory investigation; a journal of technical methods and pathology
Mariuzzi L,Domenis R,Orsaria M,Marzinotto S,Londero AP,Bulfoni M,Candotti V,Zanello A,Ballico M,Mimmi MC,Calcagno A,Marchesoni D,Di Loreto C,Beltrami AP,Cesselli D,Gri G
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Immunofluorescence
Thu Sep 01 00:00:00 EDT 2016
The regulation of cancer cell migration by lung cancer cell-derived exosomes through TGF-β and IL-10.
Oncology letters
Wang Y,Yi J,Chen X,Zhang Y,Xu M,Yang Z
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in ELISA
Mon Feb 01 00:00:00 EST 2016
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Tue Jul 19 00:00:00 EDT 2016
Follicular regulatory T cells impair follicular T helper cells in HIV and SIV infection.
Nature communications
Miles B,Miller SM,Folkvord JM,Kimball A,Chamanian M,Meditz AL,Arends T,McCarter MD,Levy DN,Rakasz EG,Skinner PJ,Connick E
Published figure using IL-10 monoclonal antibody (Product # 53-7108-42) in Flow Cytometry
Tue Oct 20 00:00:00 EDT 2015
CD4(+)CD25(-)Foxp3(+) T cells play a role in tuberculous hydrothorax rather than malignant hydrothorax.
Journal of translational medicine
Tang Y,Peng LP,Qin GX,Sun JT,Xu LJ,Jiang YF
53-7108 was used in Flow cytometry/Cell sorting to determine that CD4(+)CD25(-)Foxp3(+) T cells play a role in tuberculous hydrothorax rather than malignant hydrothorax.
Tue Aug 18 00:00:00 EDT 2015
Granzyme B production distinguishes recently activated CD8(+) memory cells from resting memory cells.
Cellular immunology
Nowacki TM,Kuerten S,Zhang W,Shive CL,Kreher CR,Boehm BO,Lehmann PV,Tary-Lehmann M
Tue May 01 00:00:00 EDT 2007
Mycobacterium bovis BCG attenuates surface expression of mature class II molecules through IL-10-dependent inhibition of cathepsin S.
Journal of immunology (Baltimore, Md. : 1950)
Sendide K,Deghmane AE,Pechkovsky D,Av-Gay Y,Talal A,Hmama Z
Sat Oct 15 00:00:00 EDT 2005