    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PSMD1 Antibodies
          Proteintech
          PSMD1 Recombinant Rabbit Monoclonal Antibody (240458C9)
          View all (5) PSMD1 antibodies
          Tech Support
          Cite PSMD1 Recombinant Rabbit Monoclonal Antibody (240458C9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          PSMD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PSMD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          PSMD1 Antibody (83521-4-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using PSMD1 antibody (83521-4-RR, Clone: 240458C9) at dilution of 1:300 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PSMD1 Antibody in Immunocytochemistry (ICC/IF)
          PSMD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PSMD1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PSMD1 Antibody in Western Blot (WB)
          PSMD1 Antibody in Flow Cytometry (Flow)
          PSMD1 Antibody in Surface plasmon resonance (SPR)
          PSMD1 Recombinant Rabbit Monoclonal Antibody (240458C9)
          Product Details
          83521-4-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:150-1:600
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240458C9
          Immunogen
          PSMD1 fusion protein Ag12009
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: AVSDVNDDVR RAAVESLGFI LFRTPEQCPS VVSLLSESYN PHVRYGAAMA LGICCAGTGN KEAINLLEPM TNDPVNYVRQ GALIASALIM IQQTEITCPK VNQFRQLYSK VINDKHDDVM AKFGAILAQG ILDAGGHNVT ISLQSRTGHT HMPSVVGVLV FTQFWFWFPL SHFLSLAYTP TCVIGLNKDL KMPKVQYKSN CKPSTFAYPA PLEVPKEKEK EKVSTAVLSI TAKAKKKEKE KEKKEEEKME VDEAEKKEEK EKKKEPEPNF QLLDNPARVM PAQLKVLTMP ETCRYQPFKP LSIGGIIILK DTSEDIEELV EPVAAHGPKI EEEEQEPEPP EPFEYIDD
          Target Information
          Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.
          Bioinformatics
          Protein Aliases: 26S proteasome non-ATPase regulatory subunit 1; 26S proteasome p112 subunit; 26S proteasome regulatory subunit RPN2; 26S proteasome regulatory subunit S1; 26S proteasome subunit p112; 26S proteasome, subunit p112; MGC133040; MGC133041; proteasome (prosome, macropain) 26S subunit, non-ATPase, 1; unnamed protein product
          View more View less
          Gene Aliases: 2410026J11Rik; P112; PSMD1; Rpn2; S1
          View more View less
          UniProt ID: (Human) Q99460, (Mouse) Q3TXS7, (Rat) O88761
          View more View less
          Entrez Gene ID: (Human) 5707, (Mouse) 70247, (Rat) 83806
          View more View less
          Function(s)
          protein binding enzyme regulator activity ubiquitin protein ligase binding protease
          Process(es)
          regulation of protein catabolic process proteasome-mediated ubiquitin-dependent protein catabolic process regulation of catalytic activity
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline