Product References
Differentiation and Characterization of Human Pluripotent Stem Cell-Derived Cardiac Endothelial Cells for In Vitro Applications.
Methods in molecular biology (Clifton, N.J.)
Raniga K,Vo NTN,Denning C
17-1449-42 was used in Flow cytometry/Cell sorting to describe a protocol for cardiac EC differentiation and a protocol for endothelial cell characterization which is are also applicable for complex model generation where multiple cardiovascular cell types are involved and may assist in optimizing differentiations for different cell lineages, including cardiomyocytes, cardiac endothelial cells, and cardiac fibroblasts.
Fri Mar 18 00:00:00 EDT 2022
Upregulation of ETV2 Expression Promotes Endothelial Differentiation of Human Dental Pulp Stem Cells.
Cell transplantation
Li J,Zhu Y,Li N,Wu T,Zheng X,Heng BC,Zou D,Xu J
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Tue Dec 21 00:00:00 EST 2021
Targeting QKI-7 in vivo restores endothelial cell function in diabetes.
Nature communications
Yang C,Eleftheriadou M,Kelaini S,Morrison T,González MV,Caines R,Edwards N,Yacoub A,Edgar K,Moez A,Ivetic A,Zampetaki A,Zeng L,Wilkinson FL,Lois N,Stitt AW,Grieve DJ,Margariti A
17-1449 was used in Flow cytometry/Cell sorting to show that Quaking-7 (QKI-7) is upregulated in iPS cell-derived endothelial cells(ECs) when exposed to hyperglycemia, and in human iPS-ECs from diabetic patients.
Thu Jul 30 00:00:00 EDT 2020
Targeting QKI-7 in vivo restores endothelial cell function in diabetes.
Nature communications
Yang C,Eleftheriadou M,Kelaini S,Morrison T,González MV,Caines R,Edwards N,Yacoub A,Edgar K,Moez A,Ivetic A,Zampetaki A,Zeng L,Wilkinson FL,Lois N,Stitt AW,Grieve DJ,Margariti A
17-1449 was used in Flow cytometry/Cell sorting to show that Quaking-7 (QKI-7) is upregulated in iPS cell-derived endothelial cells(ECs) when exposed to hyperglycemia, and in human iPS-ECs from diabetic patients.
Thu Jul 30 00:00:00 EDT 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Tue Dec 22 00:00:00 EST 2020
Angiogenic and Restorative Abilities of Human Mesenchymal Stem Cells Were Reduced Following Treatment With Serum From Diabetes Mellitus Type 2 Patients.
Journal of cellular biochemistry
Rezaie J,Mehranjani MS,Rahbarghazi R,Shariatzadeh MA
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Enhanced Therapeutic and Long-Term Dynamic Vascularization Effects of Human Pluripotent Stem Cell-Derived Endothelial Cells Encapsulated in a Nanomatrix Gel.
Circulation
Lee SJ,Sohn YD,Andukuri A,Kim S,Byun J,Han JW,Park IH,Jun HW,Yoon YS
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Tue Nov 14 00:00:00 EST 2017
Generating high-purity cardiac and endothelial derivatives from patterned mesoderm using human pluripotent stem cells.
Nature protocols
Palpant NJ,Pabon L,Friedman CE,Roberts M,Hadland B,Zaunbrecher RJ,Bernstein I,Zheng Y,Murry CE
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Endothelial Cells Promote Expansion of Long-Term Engrafting Marrow Hematopoietic Stem and Progenitor Cells in Primates.
Stem cells translational medicine
Gori JL,Butler JM,Kunar B,Poulos MG,Ginsberg M,Nolan DJ,Norgaard ZK,Adair JE,Rafii S,Kiem HP
17-1449 was used in Flow cytometry/Cell sorting to show that an ex vivo vascular niche is a platform for adult bone marrow hematopoietic stem and progenitor cell expansion.
Wed Mar 01 00:00:00 EST 2017
ERK signaling is required for VEGF-A/VEGFR2-induced differentiation of porcine adipose-derived mesenchymal stem cells into endothelial cells.
Stem cell research & therapy
Almalki SG,Agrawal DK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Fri May 12 00:00:00 EDT 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Tue Sep 12 00:00:00 EDT 2017
Changes in the pattern of plasma extracellular vesicles after severe trauma.
PloS one
Kuravi SJ,Yates CM,Foster M,Harrison P,Hazeldine J,Hampson P,Watson C,Belli A,Midwinter M,Nash GB
17-1449 was used in Flow cytometry/Cell sorting to assess the numbers of extracellular vesicles in plasma over 28 days after severe trauma and evaluate their pro-coagulant and inflammatory effects.
Mon Oct 23 00:00:00 EDT 2017
Rab11a Mediates Vascular Endothelial-Cadherin Recycling and Controls Endothelial Barrier Function.
Arteriosclerosis, thrombosis, and vascular biology
Yan Z,Wang ZG,Segev N,Hu S,Minshall RD,Dull RO,Zhang M,Malik AB,Hu G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Cellular Functions and Gene and Protein Expression Profiles in Endothelial Cells Derived from Moyamoya Disease-Specific iPS Cells.
PloS one
Hamauchi S,Shichinohe H,Uchino H,Yamaguchi S,Nakayama N,Kazumata K,Osanai T,Abumiya T,Houkin K,Era T
17-1449 was used in Flow cytometry/Cell sorting to analyze the functions and characteristics of endothelial cells in Moyamoya disease.
Wed Nov 20 00:00:00 EST 2019
Vascular niche promotes hematopoietic multipotent progenitor formation from pluripotent stem cells.
The Journal of clinical investigation
Gori JL,Butler JM,Chan YY,Chandrasekaran D,Poulos MG,Ginsberg M,Nolan DJ,Elemento O,Wood BL,Adair JE,Rafii S,Kiem HP
17-1449 was used in Flow cytometry/Cell sorting to demonstrate that endothelial Notch ligands promote PSC-definitive haematopoiesis and production of long-term engrafting CD34+ cells.
Mon Mar 02 00:00:00 EST 2015
Vascular niche promotes hematopoietic multipotent progenitor formation from pluripotent stem cells.
The Journal of clinical investigation
Gori JL,Butler JM,Chan YY,Chandrasekaran D,Poulos MG,Ginsberg M,Nolan DJ,Elemento O,Wood BL,Adair JE,Rafii S,Kiem HP
17-1449 was used in Flow cytometry/Cell sorting to demonstrate that endothelial Notch ligands promote PSC-definitive haematopoiesis and production of long-term engrafting CD34+ cells.
Mon Mar 02 00:00:00 EST 2015
Direct conversion of human amniotic cells into endothelial cells without transitioning through a pluripotent state.
Nature protocols
Ginsberg M,Schachterle W,Shido K,Rafii S
17-1449 was used in Flow cytometry/Cell sorting to develop a method to directly convert human mid-gestation lineage-committed amniotic fluid-derived cells into vascular endothelial cells.
Tue Dec 01 00:00:00 EST 2015
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Sun Feb 01 00:00:00 EST 2015
Histone Demethylases KDM4A and KDM4C Regulate Differentiation of Embryonic Stem Cells to Endothelial Cells.
Stem cell reports
Wu L,Wary KK,Revskoy S,Gao X,Tsang K,Komarova YA,Rehman J,Malik AB
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Tue Jul 14 00:00:00 EDT 2015
Distinct fibroblast lineages determine dermal architecture in skin development and repair.
Nature
Driskell RR,Lichtenberger BM,Hoste E,Kretzschmar K,Simons BD,Charalambous M,Ferron SR,Herault Y,Pavlovic G,Ferguson-Smith AC,Watt FM
17-1449 was used in Immunohistochemistry to explain why wounding is linked to formation of ECM-rich scar tissue that lacks hair follicles.
Thu Dec 12 00:00:00 EST 2013
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Fri Oct 26 00:00:00 EDT 2012
Identification of the hemogenic endothelial progenitor and its direct precursor in human pluripotent stem cell differentiation cultures.
Cell reports
Choi KD,Vodyanik MA,Togarrati PP,Suknuntha K,Kumar A,Samarjeet F,Probasco MD,Tian S,Stewart R,Thomson JA,Slukvin II
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 17-1449-42) in Flow Cytometry
Thu Sep 27 00:00:00 EDT 2012