Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD146 Antibodies

          Proteintech

          CD146/MCAM Polyclonal Antibody

          27 References
          View all (103) CD146 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD146/MCAM Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (25)
          CD146/MCAM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CD146/MCAM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 25

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD146/MCAM Antibody (17564-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed HeLa cells using 17564-1-AP (CD146/MCAM antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD146/MCAM Antibody in Immunocytochemistry (ICC/IF)
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD146/MCAM Antibody in Western Blot (WB)
          CD146/MCAM Antibody in Western Blot (WB)
          CD146/MCAM Antibody in Western Blot (WB)
          CD146/MCAM Antibody in Flow Cytometry (Flow)
          CD146/MCAM Antibody in Immunoprecipitation (IP)
          CD146/MCAM Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD146/MCAM Polyclonal Antibody

          Product Details

          17564-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:600
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 9 publications 9 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CD146/MCAM Fusion Protein Ag11762 (26-350 aa encoded by BC056418)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          60 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: GEAEQPAPE LVEVEVGSTA LLKCGLSQSQ GNLSHVDWFS VHKEKRTLIF RVRQGQGQSE PGEYEQRLSL QDRGATLALT QVTPQDERIF LCQGKRPRSQ EYRIQLRVYK APEEPNIQVN PLGIPVNSKE PEEVATCVGR NGYPIPQVIW YKNGRPLKEE KNRVHIQSSQ TVESSGLYTL QSILKAQLVK EDKDAQFYCE LNYRLPSGNH MKESREVTVP VFYPTEKVWL EVEPVGMLKE GDRVEIRCLA DGNPPPHFSI SKQNPSTREA EEETTNDNGV LVLEPARKEH SGRYECQGLD LDTMISLLSE PQELLVNYVS DVRVSP (26-350 aa encoded by BC056418)

          Target Information

          CD146, also known as MCAM (Melanoma Cell Adhesion Molecule), is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. It is heavily glycosylated, with more than 50% of its mass derived from carbohydrates. CD146 is primarily expressed on endothelial cells at cell-cell junctions, playing a crucial role in cell adhesion and the cohesion of the endothelial monolayer in vascular tissue. In addition to endothelial cells, CD146 is expressed on bone marrow fibroblasts, mesenchymal stromal cells, endometrial stromal cells, and some tumors, particularly melanoma. It has also been observed on a subset of circulating T cells and activated lymphocytes, but not on circulating endothelial cells, as indicated by the absence of other endothelial markers such as CD31 and CD51/61. CD146 functions as a calcium-independent cell adhesion molecule and may also act as a signal transduction molecule. It is involved in the recruitment of Fyn kinase and the subsequent tyrosine phosphorylation of intracellular proteins, which are important for actin cytoskeleton assembly. This signaling role includes triggering tyrosine phosphorylation of FYN and PTK2 and causing a transient increase in intracellular calcium concentration. In the context of cancer, CD146 expression may facilitate the interaction of melanoma cells with the vascular system, enhancing hematogenous tumor spread. Additionally, CD146 could serve as an adhesion molecule active in neural crest cells during embryonic development, highlighting its diverse roles in both normal physiological processes and disease states.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cell surface glycoprotein P1H12; Gicerin; l-gicerin; l-gicerin protein; melanoma adhesion molecule; melanoma-associated antigen A32; melanoma-associated antigen MUC18; P1H12 antigen; S-endo 1 endothelial-associated antigen; unnamed protein product

          View more View less

          Gene Aliases: 1-gicerin; CD146; CD149; HEMCAM; MelCAM; METCAM; MUC18; s-endo; s-gicerin

          View more View less

          UniProt ID: (Human) P43121, (Mouse) Q8R2Y2, (Rat) Q9EPF2

          View more View less

          Entrez Gene ID: (Human) 4162, (Mouse) 84004, (Rat) 78967

          View more View less

          Function(s)
          protein binding
          Process(es)
          angiogenesis glomerular filtration cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules positive regulation of cell migration vascular wound healing anatomical structure morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-86tfw:80/100.66.78.224:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0