Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 5 Antibodies

          Invitrogen

          Claudin 5 Monoclonal Antibody (4C3C2)

          Advanced Verification
          85 Published Figures
          412 References
          View all (18) Claudin 5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 5 Monoclonal Antibody (4C3C2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (85)
          • Advanced Verification (2)
          Claudin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Claudin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 90

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 5 Antibody (35-2500) in IHC (P)

          Immunohistochemistry analysis of Claudin 5 showing staining in the Membrane and Cytoplasm of paraffin-embedded human kidney tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at roo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 5 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 5 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 5 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 5 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 5 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 5 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Immunohistochemistry (IHC)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody in Western Blot (WB)
          Claudin 5 Antibody
          Claudin 5 Antibody

          Product Details

          35-2500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 174 publications 174 publications

          Immunohistochemistry (IHC)

          -
          View 110 publications 110 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          View 9 publications 9 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 15 publications 15 publications

          Immunocytochemistry (ICC/IF)

          -
          View 84 publications 84 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 14 publications 14 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Amphibian, Bovine, Chicken, Cynomolgus monkey, Dog, Guinea pig, Human, Mouse, Pig, Rat, Sheep, Xenopus, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4C3C2

          Immunogen

          Synthetic peptide derived from the mouse Claudin-5 protein

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533200

          Target Information

          This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Brain endothelial cell clone 1 protein; Claudin-5; CLDN5; Lung-specific membrane protein; TMDVCF; Transmembrane protein deleted in VCFS; transmembrane protein deleted in velocardiofacial syndrome; unnamed protein product

          View more View less

          Gene Aliases: AI854493; AWAL; BEC1; CLDN5; CPETRL1; MBEC1; TMDVCF; TMVCF

          View more View less

          UniProt ID: (Human) O00501, (Rat) Q9JKD6, (Mouse) O54942

          View more View less

          Entrez Gene ID: (Human) 7122, (Rat) 65131, (Mouse) 12741

          View more View less

          Function(s)
          structural molecule activity protein binding identical protein binding tight junction
          Process(es)
          outflow tract morphogenesis cell-cell junction assembly cell adhesion transforming growth factor beta receptor signaling pathway learning positive regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules negative regulation of angiogenesis negative regulation of cell migration maintenance of blood-brain barrier myelination negative regulation of vascular permeability response to ethanol palate development face morphogenesis bicellular tight junction assembly tight junction assembly positive regulation of establishment of endothelial barrier positive regulation of bicellular tight junction assembly establishment of blood-retinal barrier single organismal cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline