Product References
GM-CSF-activated human dendritic cells promote type 1 T follicular helper cell polarization in a CD40-dependent manner.
Journal of cell science
Korniotis S,Saichi M,Trichot C,Hoffmann C,Amblard E,Viguier A,Grondin S,Noel F,Mattoo H,Soumelis V
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Airway Administration of Bacterial Lysate OM-85 Protects Mice Against Respiratory Syncytial Virus Infection.
Frontiers in immunology
Antunes KH,Cassão G,Santos LD,Borges SG,Poppe J,Gonçalves JB,Nunes EDS,Recacho GF,Sousa VB,Da Silva GS,Mansur D,Stein RT,Pasquali C,De Souza APD
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue May 24 00:00:00 EDT 2022
The molecular and epigenetic mechanisms of innate lymphoid cell (ILC) memory and its relevance for asthma.
The Journal of experimental medicine
Verma M,Michalec L,Sripada A,McKay J,Sirohi K,Verma D,Sheth D,Martin R,Dyjack N,Seibold MA,Knapp JR,Tu TH,O'Connor BP,Gorska MM,Alam R
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
Dysregulated Peripheral Invariant Natural Killer T Cells in Plaque Psoriasis Patients.
Frontiers in cell and developmental biology
Hu Y,Chen Y,Chen Z,Zhang X,Guo C,Yu Z,Xu P,Sun L,Zhou X,Gong Y,Yu Q,Shi Y
53-9966-42 was used in Flow Cytometry, Immunocytochemistry to explore the expression of iNKT cells in psoriasis patients and the effect of secukinumab on them.
Wed Feb 23 00:00:00 EST 2022
Histone deacetylase 3 represses cholesterol efflux during CD4+ T-cell activation.
eLife
Wilfahrt D,Philips RL,Lama J,Kizerwetter M,Shapiro MJ,McCue SA,Kennedy MM,Rajcula MJ,Zeng H,Shapiro VS
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Thu Dec 02 00:00:00 EST 2021
Effector T cell responses unleashed by regulatory T cell ablation exacerbate oral squamous cell carcinoma.
Cell reports. Medicine
Chao JL,Korzinkin M,Zhavoronkov A,Ozerov IV,Walker MT,Higgins K,Lingen MW,Izumchenko E,Savage PA
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Sep 21 00:00:00 EDT 2021
LmjF.36.3850, a novel hypothetical Leishmania major protein, contributes to the infection.
Immunology
Zutshi S,Sarode AY,Ghosh SK,Jha MK,Sudan R,Kumar S,Sadhale LP,Roy S,Saha B
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
A coordinated progression of progenitor cell states initiates urinary tract development.
Nature communications
Sanchez-Ferras O,Pacis A,Sotiropoulou M,Zhang Y,Wang YC,Bourgey M,Bourque G,Ragoussis J,Bouchard M
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Immunohistochemistry
Tue May 11 00:00:00 EDT 2021
Graded RhoA GTPase Expression in Treg Cells Distinguishes Tumor Immunity From Autoimmunity.
Frontiers in immunology
Kalim KW,Yang JQ,Modur V,Nguyen P,Li Y,Zheng Y,Guo F
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Jan 11 00:00:00 EST 2022
Attenuation of TCR-induced transcription by Bach2 controls regulatory T cell differentiation and homeostasis.
Nature communications
Sidwell T,Liao Y,Garnham AL,Vasanthakumar A,Gloury R,Blume J,Teh PP,Chisanga D,Thelemann C,de Labastida Rivera F,Engwerda CR,Corcoran L,Kometani K,Kurosaki T,Smyth GK,Shi W,Kallies A
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Bendamustine with total body irradiation conditioning yields tolerant T-cells while preserving T-cell-dependent graft-versus-leukemia.
Oncoimmunology
Stokes J,Hoffman EA,Molina MS,Kummet N,Simpson RJ,Zeng Y,Katsanis E
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Wed Jul 14 00:00:00 EDT 2021
The obesity-induced adipokine sST2 exacerbates adipose Treg and ILC2 depletion and promotes insulin resistance.
Science advances
Zhao XY,Zhou L,Chen Z,Ji Y,Peng X,Qi L,Li S,Lin JD
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
Compartmentalized gut lymph node drainage dictates adaptive immune responses.
Nature
Esterházy D,Canesso MCC,Mesin L,Muller PA,de Castro TBR,Lockhart A,ElJalby M,Faria AMC,Mucida D
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
The Chemoattractant Receptor Ebi2 Drives Intranodal Naive CD4+ T Cell Peripheralization to Promote Effective Adaptive Immunity.
Immunity
Baptista AP,Gola A,Huang Y,Milanez-Almeida P,Torabi-Parizi P,Urban JF,Shapiro VS,Gerner MY,Germain RN
53-9966 was used in Flow cytometry/Cell sorting to demonstrate evolutionary selection for distinct niches within the LN that promote cellular responses, emphasizing the critical link between fine-grained tissue organization and host defense.
Tue May 21 00:00:00 EDT 2019
Characterization of Transcriptional Regulatory Networks that Promote and Restrict Identities and Functions of Intestinal Innate Lymphoid Cells.
Immunity
Pokrovskii M,Hall JA,Ochayon DE,Yi R,Chaimowitz NS,Seelamneni H,Carriero N,Watters A,Waggoner SN,Littman DR,Bonneau R,Miraldi ER
53-9966 was used in Flow cytometry/Cell sorting to study regulatory interactions between transcription factors and genes within intestinal type 1, 2, and 3 innate lymphoid cells subsets.
Tue Jul 16 00:00:00 EDT 2019
IL-33-mediated mast cell activation promotes gastric cancer through macrophage mobilization.
Nature communications
Eissmann MF,Dijkstra C,Jarnicki A,Phesse T,Brunnberg J,Poh AR,Etemadi N,Tsantikos E,Thiem S,Huntington ND,Hibbs ML,Boussioutas A,Grimbaldeston MA,Buchert M,O'Donoghue RJJ,Masson F,Ernst M
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Fri Jun 21 00:00:00 EDT 2019
TNFR2 Signaling Enhances ILC2 Survival, Function, and Induction of Airway Hyperreactivity.
Cell reports
Hurrell BP,Galle-Treger L,Jahani PS,Howard E,Helou DG,Banie H,Soroosh P,Akbari O
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Dec 24 00:00:00 EST 2019
B-Lymphocyte Phenotype Determines T-Lymphocyte Subset Differentiation in Autoimmune Diabetes.
Frontiers in immunology
Egia-Mendikute L,Arpa B,Rosell-Mases E,Corral-Pujol M,Carrascal J,Carrillo J,Mora C,Chapman H,Panosa A,Vives-Pi M,Stratmann T,Serreze D,Verdaguer J
53-9966-42 was used in Flow cytometry/Cell sorting to evaluates the T- and B-lymphocyte phenotype in two transgenic NOD mouse models were generated: the NOD-PerIg and the 116C-NOD mice.
Tue Oct 13 00:00:00 EDT 2020
Stability and function of regulatory T cells expressing the transcription factor T-bet.
Nature
Levine AG,Mendoza A,Hemmers S,Moltedo B,Niec RE,Schizas M,Hoyos BE,Putintseva EV,Chaudhry A,Dikiy S,Fujisawa S,Chudakov DM,Treuting PM,Rudensky AY
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Thu Jun 15 00:00:00 EDT 2017
Characterisation of innate lymphoid cell populations at different sites in mice with defective T cell immunity.
Wellcome open research
Dutton EE,Camelo A,Sleeman M,Herbst R,Carlesso G,Belz GT,Withers DR
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Sat Jan 09 00:00:00 EST 2021
Intranasal Administration of Lentiviral miR-135a Regulates Mast Cell and Allergen-Induced Inflammation by Targeting GATA-3.
PloS one
Deng YQ,Yang YQ,Wang SB,Li F,Liu MZ,Hua QQ,Tao ZZ
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue May 31 00:00:00 EDT 2016
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Immunohistochemistry
Fri Feb 01 00:00:00 EST 2013
Impaired proteasome function activates GATA3 in T cells and upregulates CTLA-4: relevance for Sézary syndrome.
The Journal of investigative dermatology
Gibson HM,Mishra A,Chan DV,Hake TS,Porcu P,Wong HK
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Mesenchymal stem cells alleviate TNBS-induced colitis by modulating inflammatory and autoimmune responses.
World journal of gastroenterology
Chen QQ,Yan L,Wang CZ,Wang WH,Shi H,Su BB,Zeng QH,Du HT,Wan J
Published figure using Gata-3 monoclonal antibody (Product # 53-9966-42) in Western Blot
Wed Aug 07 00:00:00 EDT 2013
Role of CD44 in the differentiation of Th1 and Th2 cells: CD44-deficiency enhances the development of Th2 effectors in response to sheep RBC and chicken ovalbumin.
Journal of immunology (Baltimore, Md. : 1950)
Guan H,Nagarkatti PS,Nagarkatti M
Wed Jul 01 00:00:00 EDT 2009