    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • mCherry Antibodies
          Invitrogen
          mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 594
          13 Published Figures
          10 References
          View all (35) mCherry antibodies
          Tech Support
          Cite mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 594
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (13)
          mCherry Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          mCherry Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 14
          mCherry Antibody (M11240) in Flow
          Samples were acquired and analyzed using 561 nm excitation specified band pass emission filter on a BD LSR II flow cytometer. The histogram shows cells stained with mCherry Rat Monoclonal Ab Alexa Fluor® 594 Conjugate. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          mCherry Antibody in Flow Cytometry (Flow)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          mCherry Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          mCherry Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          M11240
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 1 publication 1 publication
          Immunohistochemistry (Frozen) (IHC (F))
          -
          View 1 publication 1 publication
          Immunohistochemistry - Free Floating (IHC (Free))
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          -
          View 4 publications 4 publications
          Flow Cytometry (Flow)
          Assay-dependent
          View 3 publications 3 publications
          Product Specifications
          Species Reactivity
          Tag
          Published species
          Tag
          Host/Isotype
          Rat / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          16D7
          Immunogen
          Full-length mCherry was used as the immunogen
          Conjugate
          Alexa Fluor™ 594 Alexa Fluor™ 594 Alexa Fluor™ 594
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 647 
          • Pacific Blue
          • Request custom conjugation
          Excitation/Emission Max
          590/618 nm View spectra spectra
          Form
          Liquid
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.5
          Contains
          5mM sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536614
          Target Information
          MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
          Bioinformatics
          Protein Aliases: mCherry
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-5d76f9f486-n5jxz:80/100.66.73.31:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod