    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • SSEA3 Antibodies
          Invitrogen
          SSEA3 Monoclonal Antibody (MC-631), DyLight™ 488
          2 References
          View all (10) SSEA3 antibodies
          Tech Support
          Cite SSEA3 Monoclonal Antibody (MC-631), DyLight™ 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          SSEA3 Antibody (MA1-020-D488) in ICC/IF
          Immunofluorescent analysis of SSEA-3 (green) showing membrane staining of NCCIT cells (right panel) compared to negative HeLa cell control (left panel). The cells were fixed with formalin for 15 minutes, washed, and then blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a DyLight 488-conjugated SSEA-3 monoclonal antibody (Product # MA1-020-D488) in 3% BSA-PBS at a dilution of 1:20 and incubated for 1 hour at 37C in the dark. F-Actin (left panel, red) was stained with DyLight 554 Phalloidin (Product # 21834)... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Immunocytochemistry (ICC/IF)
          SSEA3 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA1-020-D488
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:10-1:100
          -
          Flow Cytometry (Flow)
          1:50-1:500
          View 2 publications 2 publications
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Published species
          Human, Sheep
          Host/Isotype
          Rat / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MC-631
          Immunogen
          Fischer rat immunized with 4-8 cell stage mouse embryos
          Conjugate
          DyLight™ 488 DyLight™ 488 DyLight™ 488
          • Unconjugated
          • DyLight 650
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          492/519 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography - MBP
          Storage buffer
          PBS with proprietary stabilizer
          Contains
          0.02% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536683
          Product Specific Information
          MA1-020-D488 has been successfully used in ICC/IF and flow cytometry applications on human samples.
          MA1-020-D488 can be used for immunofluoresence analysis of SSEA3 in human iPSCs.
          Target Information
          SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: SSEA-3; stage-specific embryonic antigen 3
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-vjw2z:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline