    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SSEA5 Antibodies
          Invitrogen
          SSEA5 Monoclonal Antibody (8E11), DyLight™ 488
          View all (4) SSEA5 antibodies
          Questions & Answers
          Cite SSEA5 Monoclonal Antibody (8E11), DyLight™ 488
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          SSEA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SSEA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          SSEA5 Antibody (MA1-144-D488) in ICC/IF
          Immunofluorescent analysis of SSEA-5 (green) showing membrane staining of NCCIT cells (right panel) compared to negative HeLa control (left panel). The cells were fixed with formalin for 15 minutes, washed, and then blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a DyLight 488-conjugated SSEA-5 monoclonal antibody (Product # MA1-144-D488) in 3% BSA-PBS at a dilution of 1:20 and incubated for 1 hour at 37C in the dark. F-Actin (left panel, red) was stained with DyLight 554 Phalloidin (Product # 21834) and ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SSEA5 Antibody in Immunocytochemistry (ICC/IF)
          SSEA5 Antibody in Immunocytochemistry (ICC/IF)
          SSEA5 Antibody in Immunocytochemistry (ICC/IF)
          SSEA5 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA1-144-D488
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:10-1:100
          -
          Flow Cytometry (Flow)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          8E11
          Immunogen
          Undifferentiated H9 human embryonic stem cells
          Conjugate
          DyLight™ 488 DyLight™ 488 DyLight™ 488
          • Unconjugated
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          492/519 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS with proprietary stabilizer
          Contains
          0.02% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536849
          Product Specific Information
          Immunofluorescence and flow cytometric analysis of MA1-144-D488 shows positive staining of SSEA-5 positive pluripotent cells and cultured human embryonal carcinoma NCCIT cells.
          Target Information
          Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: SSEA-5
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0