    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HuC/HuD Antibodies
          Invitrogen
          HuC/HuD Recombinant Rabbit Monoclonal Antibody (K01_1S63)
          View all (2) HuC/HuD antibodies
          Questions & Answers
          Cite HuC/HuD Recombinant Rabbit Monoclonal Antibody (K01_1S63)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HuC/HuD Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HuC/HuD Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HuC/HuD Antibody (MA567856) in WB
          Western Blot analysis of HuC/HuD in rat Brain lysates using HuC/HuD Recombinant Monoclonal Antibody (Product # MA567856). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HuC/HuD Antibody in Western Blot (WB)
          Product Details
          MA567856
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_1S63
          Immunogen
          Recombinant protein of mouse HuC.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 0.05% BSA, 40% glycerol, 150mM NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Anti-Hu antibodies were originally isolated from a patient with paraneoplastic encephalomyelitis, a rare condition of selective neural tissue injury associated with small cell lung carcinoma. The antibodies bind specifically to antigens present exclusively in neuronal cells and are thus useful as markers of neuronal cells in tissue. The Hu antigen is an RNA-binding protein of the embryonic lethal abnormal visual (Elav) family.
          Bioinformatics
          Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV-like protein 3; ELAV-like protein 4; Hu-antigen D; HuD; Paraneoplastic encephalomyelitis antigen HuD
          View more View less
          Gene Aliases: Elavl4; Huc; HuD; r-HuD; RGD1561943
          View more View less
          UniProt ID: (Rat) O09032
          View more View less
          Entrez Gene ID: (Rat) 282824, (Rat) 432358
          View more View less
          Function(s)
          nucleotide binding AU-rich element binding mRNA binding mRNA 3'-UTR binding protein binding translation initiation factor
          Process(es)
          nervous system development aging learning locomotory behavior associative learning neuron differentiation regeneration response to endoplasmic reticulum stress response to cocaine regulation of mRNA stability dendrite morphogenesis positive regulation of dendrite development cellular response to nerve growth factor stimulus
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline