Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RASA3 Antibodies

          Abnova

          RASA3 Polyclonal Antibody, MaxPab™

          Greener Choice
          View all (11) RASA3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RASA3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          RASA3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RASA3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RASA3 Antibody (H00022821-D01P) in WB

          Western Blot analysis of RASA3 expression in transfected 293T cell line (H00022821-T02) by RASA3 MaxPab polyclonal antibody.

          Lane 1: RASA3 transfected lysate(95.70 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RASA3 Antibody in Western Blot (WB)
          RASA3 Antibody in Western Blot (WB)
          RASA3 Polyclonal Antibody, MaxPab™

          Product Details

          H00022821-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RASA3 (NP_031394.2, 1 a.a. approximately 834 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAVEDEGLRV FQSVKIKIGE AKNLPSYPGP SKMRDCYCTV NLDQEEVFRT KIVEKSLCPF YGEDFYCEIP RSFRHLSFYI FDRDVFRRDS IIGKVAIQKE DLQKYHNRDT WFQLQHVDAD SEVQGKVHLE LRLSEVITDT GVVCHKLATR IVECQGLPIV NGQCDPYATV TLAGPFRSEA KKTKVKRKTN NPQFDEVFYF EVTRPCSYSK KSHFDFEEED VDKLEIRVDL WNASNLKFGD EFLGELRIPL KVLRQSSSYE AWYFLQPRDN GSKSLKPDDL GSLRLNVVYT EDHVFSSDYY SPLRDLLLKS ADVEPVSASA AHILGEVCRE KQEAAVPLVR LFLHYGRVVP FISAIASAEV KRTQDPNTIF RGNSLASKCI DETMKLAGMH YLHVTLKPAI EEICQSHKPC EIDPVKLKDG ENLENNMENL RQYVDRVFHA ITESGVSCPT VMCDIFFSLR EAAAKRFQDD PDVRYTAVSS FIFLRFFAPA ILSPNLFQLT PHHTDPQTSR TLTLISKTVQ TLGSLSKSKS ASFKESYMAT FYEFFNEQKY ADAVKNFLDL ISSSGRRDPK SVEQPIVLKE GFMIKRAQGR KRFGMKNFKK RWFRLTNHEF TYHKSKGDQP LYSIPIENIL AVEKLEEESF KMKNMFQVIQ PERALYIQAN NCVEAKDWID ILTKVSQCNQ KRLTVYHPSA YLSGHWLCCR APSDSAPGCS PCTGGLPANI QLDIDGDRET ERIYSLFNLY MSKLEKMQEA CGSKSVYDGP EQEEYSTFVI DDPQETYKTL KQVIAGVGAL EQEHAQYKRD KFKKTKYGSQ EHPIGDKSFQ NYIRQQSETS THSI

          Target Information

          Human GAP1-InsP4 BP, also designated Ras p21 protein activator (GTPase-activating protein) 3 [Ins(1,3,4,5)P4-binding protein], is an 829-amino acid protein that binds phospholipids in both a calcium-dependent and -independent manner. GAP1, one of the Ras GTPase-activating protein families, comprises four distinct genes, including GAP1(m), GAP1-InsP4 BP, MRASAL (murine Ras GTPase-activating-like) and KIAA0538. This family contains an N-terminal tandem C2 domain, a GAP-related domain and a C-terminal pleckstrin homology (PH) domain. The PH domains of GAP1-InsP4 BP are essential for membrane targeting via binding of specific phospholipids. Following agonist-stimulated PtdIns(3,4,5)P(3) production, group I family PH domain containing proteins like GAP1-InsP4 BP specifically bind inositol phosphates, which are subsequently targeted to the plasma membrane.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GAP1(IP4BP); GAP1-InsP4 BP; GapIII; GAPIII activator 3; GTPase activating protein III; GTPase-activating protein 1 family, inositol 1,3,4,5-tetrakisphosphate-binding protein; Ins P4-binding protein; Ins(1,3,4,5)P4-binding protein; R-Ras gap; R-ras RTPase activating protein-3; Ras GTPase-activating protein 3; Ras GTPase-activating protein III; RP11-245B11.3; unnamed protein product

          View more View less

          Gene Aliases: AI326412; C86362; E130011G04; GAP1IP4BP; GAPIII; RASA3; scat

          View more View less

          UniProt ID: (Human) Q14644, (Mouse) Q60790

          View more View less

          Entrez Gene ID: (Human) 22821, (Mouse) 19414

          View more View less

          Function(s)
          GTPase activator activity calcium channel regulator activity zinc ion binding metal ion binding calcium-release channel activity GTPase-activating protein
          Process(es)
          response to stress signal transduction platelet activation intracellular signal transduction negative regulation of Ras protein signal transduction regulation of intracellular signal transduction cellular response to heat regulation of GTPase activity positive regulation of GTPase activity calcium ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline