Phenom ProSuite Desktop SEM Software
The Thermo Scientific ProSuite Software Package enables Phenom users to extract maximum information from images made with the Thermo Scientific Phenom Desktop Scanning Electron Microscope (SEM). It extends the capabilities of the high-resolution Phenom Desktop SEM in order to satisfy specific application needs. ProSuite Software is installed directly on the Phenom workstation.
The Phenom Application System and ProSuite Software are available for any Phenom Desktop SEM. The Phenom Application System can be connected directly via a local network or the internet, enabling network storage and remote system control. 
Key Features
Automated image mapping
The automated image mapping application enables users to create large field-of-view SEM images with high resolution.
Remote user interface
The remote user interface of the Phenom ProSuite Software makes it possible to access the Phenom Desktop SEM from a different location.
Time-lapse recorder
Allows you to create time-lapse recordings from SEM images.
Specifications
Hoja de estilo para las especificaciones de la tabla de productos
  • Automated collection of images
  • Real-time remote control
  • Intuitive single-page user interface 
  • Standard applications included: automated image mapping and remote user interface
  • Dedicated for all Phenom Desktop SEM
  • Large field of view (FOV) images (min. magnification 31.8x, max. FOV 8.07 mm)
Style Sheet for Komodo Tabs
Resources
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.