FoamAway Irradiated AOF
Gibco FoamAway Irradiated AOF is an animal origin-free antifoaming/defoaming agent for use in cell culture systems. FoamAway product has a sterility assurance of 10-6 and is available in ready-to-use packaging or customized for your specific application.
Order now
Bag size and fill volume
Bag size | Fill volume (L) | Fill volume (% of bag size) | Standard increments |
---|---|---|---|
250mL | 0.25 | 100% | N/A |
1L | 0.3 - 1 | 30 - 100% | 10% or 100mL |
5L | 1.5 - 5 | 30 - 100% | 10% or 500mL |
10L | 4 - 10 | 40 - 100% | 10% or 1L |
20L | 6 - 10 | 30 - 50% | 10% or 2L |
Option A. 250mL / 1L bag with 3 base ports
Tube diameter (ID x OD) |
Tube length | Tube type | Connector type | |
---|---|---|---|---|
Base Port 1 | Fill port for internal use only | |||
Base Port 2 | 1/4" ID | 3" | PVC | Spike port |
Base Port 3 | 1/4"x3/8" | 3" | C-flex | Female luer adapter, male septum cap |
Option B: 5L / 10L / 20L bag with 4 base ports
Tube diameter (ID x OD) |
Tube length | Tube type | Connector type | ||||
---|---|---|---|---|---|---|---|
Base Port 1 | 1/8" x 1/4" | 40" | C-flex | Female luer adapter | |||
Base Port 2 | 3/8" x 5/8" | 40" | C-flex | MPC quick connect | |||
Base Port 3 | 1/4" ID | 3" | PVC | Spike port | |||
Base Port 4 | 3/16" x 5/16" | 40" | C-flex | Male luer adapter, female cap |
Tube Diameter, Material and Connector Options
Tube diameter (ID x OD) |
Tube material options | Connector options |
---|---|---|
.125" x .255" | C-flex, Pharmed, Silicone | Threaded male and female luers |
1/8" x 1/4" | C-flex, Pharmed, Silicone | Threaded male and female luers |
.151" x .215" | PVC | Threaded male and female luers |
3/16" x 5/16" | C-flex, Pharmed, Silicone | Threaded male and female luers; blood spikes |
1/4" x 7/16" | C-flex, PVC, Silicone | Threaded male and female luers; MPC quick connects; blood spikes; Tri-clover clamps |
3/8" x 5/8" | C-flex, Pharmed, Silicone | All MPC quick connects; Kleenpak |
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.