Search
The Thermo Scientific Cytomat Wide Range Series provides robot-friendly random access storage of up to 1512 standard microplates and can be easily integrated into any automated system for multiple applications. The series accommodates a range of temperatures (4°C to 70°C) and humidities, as well as gaseous atmospheres.
| Cytomat 2 C470-LiN | Incubation/storage for cell culture applications from 4 to 70°C. Humidity up to 95% r.H., CO2 range of 0-20 Vol. %, ContraCon decontamination routine. |
| Cytomat 2 C450-LiN | Incubation/storage for cell culture applications 4 to 50°C, humidity up to 95% r.H., and CO2 Range of 0-20 Vol. %, ContraCon decontamination routine. |
| Cytomat 5 C450 | Midsize automated incubator and storage solution for up to 105 (126) standard microplates with dependable sample protection, a temperature range of +4°C to 50°C, controlled humidity up to 95% r.H., and stable CO2 conditions. |
| Cytomat 10 C450 | Fastest, high-capacity incubation and storage for up to 210 standard microplates with dependable sample protection, a temperature range of 4°C to 50°C, and stable CO2 conditions. |
| Cytomat 24 C450 | Ideal for highest capacity storage and incubation of up to 504 standard microplates, a temperature range of +4°C to 50°C, humidity up to 95% r.H., and stable CO2 conditions. |
| Cytomat 48 C450 | Two Cytomat 24 C450 incubators combined into one to accommodate up to 1008 standard microplates, a temperature range of +4°C to 50°C, humidity up to 95% r.H., and stable CO2 conditions. |
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation.
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
This swap station accelerates your lab automation handling system by providing two plate positions.
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke.
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more.
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Many more options available
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation.
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
This swap station accelerates your lab automation handling system by providing two plate positions.
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke.
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more.
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Many more options available