Electrophoresis with Sanger Sequencing
During capillary electrophoresis, the products of the cycle sequencing reaction are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the negatively charged DNA fragments move through the polymer in the capillaries toward the positive electrode.
A high voltage is applied so that the negatively charged DNA fragments move through the polymer in the capillaries toward the positive electrode (Figure 1). Capillary electrophoresis can resolve DNA molecules that differ in molecular weight by only one nucleotide.
Shortly before reaching the positive electrode, the fluorescently labeled DNA fragments, separated by size, move through the path of a laser beam. The laser beam causes the dyes on the fragments to fluoresce. An optical detection device on Applied Biosystems DNA analyzers detects the fluorescence (Figure 2).
The Data Collection Software converts the fluorescence signal to digital data, then records the data in a *.ab1 file. Because each dye emits light at a different wavelength when excited by the laser, all four colors, and therefore, all four bases, can be detected and distinguished in one capillary injection.
Capillary Electrophoresis instrument selection guide
![]() 3730xL |
![]() 3730 |
![]() 3500xL |
![]() 3500 |
![]() 3130xL |
![]() 3130 |
![]() 310 |
|
Instrument | |||||||
---|---|---|---|---|---|---|---|
Number of Capillaries | 96 | 48 | 24 | 8 | 16 | 4 | 1 |
Compatible Applications: (S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported |
|||||||
BAC End Sequencing | S | S | S | S | S | S | S |
Checking Clone Constructs | S | S | S | S | S | S | S |
de novo Sequencing | S | S | S | S | S | S | S |
Heterozygote Detection & Resequenicng (SNP-based) | S | S | S | S | S | S | A |
HLA Typing | N | N | S | S | S | S | C |
Methylation | S | S | S | S | S | S | C |
mtDNA Sequencing | S | S | S | S | S | S | S |
Resequencing | |||||||
- Comparative Genomics | S | S | S | S | S | S | S |
- Indels | S | S | S | S | S | S | S |
SAGE™ Method | C | C | C | C | C | C | A |
SNP Analysis | S | S | S | S | S | S | S |
General Sizing of PCR Products | S | S | S | S | S | S | S |
Fragment Length Polymorphism | |||||||
- AFLP | S | S | S | S | S | S | S |
- RFLP | A | A | A | A | A | A | A |
Large Fragment Analysis | |||||||
- BAC Fingerprinting | A | A | A | A | A | A | N |
- VNTR | C | C | C | C | C | C | C |
Microsatellite | |||||||
- Genotyping | S | S | S | S | S | S | S |
- Analysis for Forensics/HID | N | N | S | S | S | S | S |
- Instability/RER | A | A | A | A | A | A | A |
Chimerism Studies | C | C | C | C | C | C | C |
Conformation Analysis | |||||||
- SSCP | N | N | N | N | A | A | S |
Relative Fluorescent Quantitation | |||||||
- Loss of Heterozygosity (LOH) | S | S | S | S | S | S | S |
SNP Genotyping | |||||||
- SNaPshot® System | A | A | S | S | S | S | S |
- SNPlex™ System | S | S | S | S | S | N | N |
DNA Protein Binding Assays & DNA Fingerprinting | C | C | C | C | C | C | C |
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
For Research Use Only. Not for use in diagnostic procedures.