Search
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
Wipe the magnetic rods with a soft cloth or tissue paper soaked in a mild detergent solution, soap solution, or alcohol.
The protocol will not start without the tip combs inserted into the tip comb holder.
If the starting material is too viscous, the magnetic rods will not be able to collect the particles. Dilute the sample and check that the sample is properly homogenized and lysed.
This happens sometimes, but it will not affect the yield because the sample has been released from the particles.
A potential cause is that the plastic tip comb has warped slightly due to its design. This is why they are packed in pairs, so that they maintain their shape. Flex the backbone of the tip comb and try again.
Turn the machine off, and inspect it for any visible damage. Make sure the correct head is being used for the protocol being run. If the machine shows no visible damage, turn the machine back on. This should reset the machine. If the problem immediately occurs again, turn the machine off and gently move the magnetic head apparatus to the center of its travel path, then turn the machine on again. If the problem persists, the head may need to be realigned by a service call.
There is a pause function if you hit "stop" in the middle of a run. If you hit start again, the run continues. If you hit stop twice, the run stops completely. There is no way to begin a run in the middle of a protocol.
Unfortunately, freezing of the RNA binding beads will render them nonfunctional. Please discard them.
At lower temperatures, the components of the Lysis/Binding Solution and Wash I Solution may precipitate. To get rid of the precipitate, just warm the solutions to room temperature and invert bottles gently to solubilize the reagents before use.
Wipe the magnetic rods with a soft cloth or tissue paper soaked in a mild detergent solution, soap solution, or alcohol.
The protocol will not start without the tip combs inserted into the tip comb holder.
If the starting material is too viscous, the magnetic rods will not be able to collect the particles. Dilute the sample and check that the sample is properly homogenized/lysed. Adding low amounts of detergent will improve the collection of magnetic particles as well. Quickly centrifuge the plate to sediment the particles to the bottom of the plate.
This happens sometimes but it will not affect the yield because the sample has been released from the particles.
No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand or a modified version of the original KingFisher Flex protocol would have to be created that begins at the point where the run was stopped.
Please follow these recommended steps:
For Research Use Only. Not for use in diagnostic procedures.