Search
The global prevalence of sexually transmitted diseases (STIs) is over 1 million cases acquired every day, most of which are asymptomatic. There are approximately 374 million new infections each year, including three of the most common curable STIs: chlamydia, gonorrhea, and trichomoniasis.* These three STIs, as well as mycoplasma genitalium, can lead to complications including cervicitis, urethritis, preterm labor, and pelvic inflammatory disease (PID), and can increase the risk of contracting HIV. Thus, the detection and research of common prevalent STIs is of critical importance for clinical research to understand outbreaks and disease etiology.
The Applied Biosystems TrueMark STI Select Panel employs real-time PCR technology, offering an end-to-end workflow for the detection of common sexually transmitted infections by utilizing the power of Applied Biosystems QuantStudio real-time PCR systems.**
The TrueMark STI Select Panel includes multiplexed assays in a single-tube format to detect chlamydia trachomatis, neisseria gonorrhoeae, mycoplasma genitalium, and trichomonas vaginalis, as well as RNase P as an internal process control in the same reaction. Utilizing a QuantStudio system enables rapid, accurate detection and categorization of microorganisms that cause these common sexually transmitted diseases.
TrueMark STI Select Panel
Product details |
|
TrueMark STI Select Panel, Combo Kit components | TrueMark STI Select Panel TrueMark Infectious Disease 1-Step Multiplex Master Mix (No ROX) |
Targets | Chlamydia trachomatis (CT) Neisseria gonorrhoeae (NG) Mycoplasma genitalium (MG) Trichomonas vaginalis (TV) |
Internal process control | RNase P |
Positive control | TrueMark STI Amplification Control*** |
Sample type | Urine†, vaginal† or genital swab |
Real-time PCR instruments | • QuantStudio 5 Real-Time PCR System (96-well, 0.2 mL) • QuantStudio 5 Dx Real-Time PCR System (96-well, 0.2 mL)** • QuantStudio 7 Pro Real-Time PCR System • QuantStudio 7 Pro Dx Real-Time PCR System** • QuantStudio 7 Flex Real-Time PCR System • QuantStudio 12 Flex Real-Time PCR System |
Turnaround time | Less than 4 hours |
Software | QuantStudio Design and Analysis v2.6 or later |
**QuantStudio 5 Dx and QuantStudio 7 Pro Dx systems are For In Vitro Diagnostic Use. Test development mode is for Research Use Only. Not for use in diagnostic procedures.
***TrueMark STI Amplification Control required. Sold separately.
†Internally tested sample type.
We offer more STI and vaginal infection research testing solutions, in predefined panels that include 8-targets each or create your own custom panel from a large collection of microbial and antibiotic resistance assays , all prespotted on TaqMan Array Plates
Implementing genetic analysis systems and solutions in the laboratory supports improved efficiencies, cost savings, and faster time to reportable results. qPCR can detect slow-growing, difficult-to-cultivate microorganisms, making it ideal for when culture methods are inadequate, ambiguous, time-consuming, difficult, or costly.[1,2]
Learn more about implementing molecular testing with qPCR in your lab ›
Utilize an end-to-end workflow to expand your molecular infectious disease research menu, using the TrueMark STI Select Panel, a single-tube multiplexed assay option including four STI targets and an internal process control, optimized for use on Quantstudio qPCR systems. Go from sample to result in approximately four hours.
New to Thermo Fisher molecular testing? Easily click on the steps to below to see the recommended components needed to set up your laboratory for real-time PCR testing.
†Internally tested sample type.
Utilize an end-to-end workflow including KingFisher systems and MagMAX kits
![]() | ![]() | ![]() | ![]() |
|
|
|
|
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
|
|
|
|
|
|
QuantStudio 5 Dx and QuantStudio 7 Pro Dx systems are For In Vitro Diagnostic Use. Test development mode is for Research Use Only. Not for use in diagnostic procedures.
†Internally tested sample type.
Utilize an end-to-end workflow including KingFisher systems and MagMAX kits
![]() | ![]() | ![]() | ![]() |
|
|
|
|
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
|
|
|
|
|
|
QuantStudio 5 Dx and QuantStudio 7 Pro Dx systems are For In Vitro Diagnostic Use. Test development mode is for Research Use Only. Not for use in diagnostic procedures.
References:
1. Finegold, David N. “Genetic Diagnostic Technologies - Fundamentals.” Merck Manuals Consumer Version, Merck Manuals, 21 Feb. 2023, https://www.merckmanuals.com/home/fundamentals/genetics/genetic-diagnostic-technologies.
2. Vazquez-Pertejo, Maria T. “Diagnosis of Infectious Disease - Infections.” Merck Manuals Consumer Version, Merck Manuals, 21 Feb. 2023, https://www.merckmanuals.com/home/infections/diagnosis-of-infectious-disease/diagnosis-of-infectious-disease.
*Sexually transmitted infections (STIs) (2022) World Health Organization. Available at: https://www.who.int/news-room/fact-sheets/detail/sexually-transmitted-infections-(stis).
**QuantStudio 5 Dx and QuantStudio 7 Pro Dx systems are For In Vitro Diagnostic Use. Test development mode is for Research Use Only. Not for use in diagnostic procedures.
***TrueMark STI Amplification Control required. Sold separately.
†Internally tested sample type.
For Research Use Only. Not for use in diagnostic procedures.
© 2023 Thermo Fisher Scientific Inc. All rights reserved.
All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.