Comprehensive guide to selecting the right web gauge for thickness and basis weight measurement
Use this comparison chart to see which system is right for you.
Applications
21PlusHD
Iplus!
Cast film
Co-extruded film
Sheet extrusion
Biax film
Converted products
Extrusion coating (total only)
Extrusion coating (base/coat/total)
Tandem extrusion coating
Roll/blade coating
Building materials
Paper
Nonwovens
Patch coating
Vinyl calendering
Roofing
Abrasives
Scanner/sensor compatibility chart
Integra
Mark III
C-Frame
Box Beam
PROSIS IR Transmission
PROSIS IR Reflection
PROSIS IR Dual Reflection
Beta Plus HE
✓*
✓*
PM Roofing Sensor
Xray Plus
✓*
ShadowMaster
*Available on Iplus! platform
Sensors
21PlusHD
Iplus!
Range
Measurement
Measurement Principle
Beta Plus HE KR85
5–1600 gsm
Basis weight
Isotope
BetaPlus SR90
100–7000 gsm
Basis weight
Isotope
PM Roofing  SR90
100–7000 gsm
Basis weight
Isotope
Prosis
1–2000 µm depending on material
Basis weight or direct thickness
NIR spectroscopy
ShadowMaster
100–6500 µm
Direct thickness
Laser
Xray Plus
20–50,000 gsm
Basis weight
X-ray
Scanner/sensor compatibility chart
Integra
Mark III
C-Frame
Box Beam
PROSIS IR Transmission
PROSIS IR Reflection
PROSIS IR Dual Reflection
Beta Plus HE
✓*
✓*
PM Roofing Sensor
Xray Plus
✓*
ShadowMaster
*Available on Iplus! platform
Scanners
21Plus!
Iplus!
Max Sheet Width
Mark III Industrial Scanner
✓*
393.7 in. (10m)
Mark III Hazard Reduced Scanner
148 in. (3749mm)
Mark III Industrial Scanner for ShadowMaster Sensor (includes roll)
118 in. (2997mm)
Mark III Vertical Passline Industrial Scanner—CAN BE TILTED +/- 45 DEGREES FROM VERTICAL
148 in. (3749mm)
Mark III Wide Gap Industrial Scanner 
148 in. (3749mm)
Abrasive Scanning C-Frame—ALWAYS VERTICAL
80 in. (2032mm)
Stainless Steel Roofing Scanning (High Moisture Content) C-Frame
80 in. (2032mm)
Integra Scanner
88 in. (2236mm)
Box Beam
80 in. (2032mm)
*Mark III with iPlus! Platform available up to 160 in (4064mm)
Scanner/sensor compatibility chart
Integra
Mark III
C-Frame
Box Beam
PROSIS IR Transmission
✓*
PROSIS IR Microbeam
PROSIS IR Reflection
PROSIS IR Dual Reflection
Beta Plus HE
✓*
✓*
PM Roofing Sensor
Xray Plus
ShadowMaster
*Available on Iplus! platform
For Research Use Only. Not for use in diagnostic procedures.