Search
Fluorescent lectins have been used to localize and detect cell-surface or intracellular glycoconjugates, making them versatile primary detection reagents for microscopy or with flow cytometry. Lectins are available conjugated to a wide variety of fluorescent dyes, including bright, photostable Alexa Fluor dyes, increasing the flexibility in designing multicolor experiments for imaging and flow cytometry. Special conjugation procedures are used to optimize the number of dye molecules bound to the lectins, leaving a bright signal while preserving the binding specificity of the individual lectin. High signal-to-noise ratio is also maintained by the removal of unconjugated fluorochromes and inactive lectins.
Lectins are carbohydrate binding proteins that are highly specific to oligosaccharides, or sugar groups attached to other molecules. They can cause agglutination of specific cells or precipitation of glycoconjugates. Lectins are involved in regulation of cell adhesion, glycoprotein synthesis, and blood protein levels along with playing a role in the immune system.
Oligosaccharides are found throughout the cell surface but can also be found attached within the cell. Specific oligosaccharides and their distribution can vary among cell type and organelle. Lectins, because of their specificity to their target, can serve to identify cell types or cellular components when used in conjunction with various labeling approaches. Fluorescent derivatives, or conjugated lectins, have been used to localize and detect cell-surface or intracellular glycoconjugates, making them versatile primary detection reagents for microscopy or with flow cytometry. Additionally, cellular differentiation of cancerous cell types are often identified by the makeup of their surface glycoproteins, making lectins useful markers in detecting those distinct cellular phenotypes.
| Lectin | Conjugate | Source | Preferred sugar specificity | Blood group specificity | Mitogenic activity |
|---|---|---|---|---|---|
| Arachis hypogaea (peanut) lectin (PNA) | Alexa Fluor 488 | Peanut | T antigen (M, N) | ß-gal(1->3)galNAc | No |
| Alexa Fluor 568 | |||||
| Alexa Fluor 594 | |||||
| Alexa Fluor 647 | |||||
| Concanavalin A (Con A) | Alexa Fluor 350 | Canavalia ensiformis (jack bean) | Non-specific after neuraminidase treatment | α-man, α-glc | Yes |
| Alexa Fluor 488 | |||||
| Alexa Fluor 594 | |||||
| Alexa Fluor 633 | |||||
| Alexa Fluor 647 | |||||
| Texas-Red | |||||
| Tetramethylrhodamine (TRITC) | |||||
| Concanavalin A (ConA), succinylated | Alexa Fluor 488 | Canavalia ensiformis (jack bean) | Non-specific after neuraminidase treatment | α-man, α-glc | Yes |
| Dolichos biflorus agglutinin (DBA) | Fluorescein (FITC) | Horse gram | A1 | α-galNAc | No |
| Glycine max (soybean) lectin (SBA) | Alexa Fluor 594 | Soybean | A>O>B | α>ßGalNAc | Yes |
| Alexa Fluor 647 | |||||
| Griffonia simplicifolia lectin (GSL-II) | Alexa Fluor 488 | Africa shrub legume | B>>A1 | α or ß GalNAc | No |
| Alexa Fluor 594 | |||||
| Alexa Fluor 647 | |||||
| Griffonia simplicifolia isolectin GS-IB4 of B subunits (GS-IB4, GSL-IB4) | Alexa Fluor 488 | Africa shrub legume | B | αGal | No |
| Alexa Fluor 568 | |||||
| Alexa Fluor 594 | |||||
| DyLight 594 | |||||
| Alexa Fluor 647 | |||||
| Biotin-XX conjugate | |||||
| Helix pomatia lectin (HPA) | Alexa Fluor 488 | Edible snail | A | galNAc | No |
| Alexa Fluor 647 | |||||
| Lens culinaris agglutinin (LCA) | Fluorescein (FITC) | Lentil | Non-specific after neuraminidase treatment | αMan, αGlc | Yes |
| Lotus tetragonolobus lectin (LTL) | Fluorescein (FITC) | Asparagus pea | O<A2 | αFuc | Yes |
| Lycopersicon esculentum lectin (LEL, LEA, TL) | DyLight 488 | Tomato | Non-specific after neuraminidase treatment | (glcNAc)3 | Yes |
| DyLight 594 | |||||
| DyLight 649 | |||||
| Fluorescein (FITC) | |||||
| Texas Red | |||||
| Phaseolus vulgaris lectin (PHA-L) | Alexa Fluor 488 | Red kidney bean | Non-specific after neuraminidase treatment | Galß4GlcNAcß6(GlcNAc ß2Manα3)Manα3 | Yes |
| Alexa Fluor 594 | |||||
| Ricinus communis agglutinin I (RCA I, RCA 120) | Fluorescein (FITC) | Castor bean | Non-specific after neuraminidase treatment | Gal | No |
| Sambucus nigra lectin (SNA, EBL) | Fluorescein (FITC) | Elderberry | Non-specific after neuraminidase treatment | Neu5Acα6Gal/GalNAc | No |
| Ulex europaeus agglutinin I (UEA I) | Fluorescein (FITC) | Gorse, furze | o>A2 | αFuc | No |
| Wheat germ agglutinin (WGA) | Alexa Fluor 350 | Wheat germ | A, B, O | GlcNAc | Yes |
| Alexa Fluor 488 | |||||
| Alexa Fluor 555 | |||||
| Alexa Fluor 594 | |||||
| Alexa Fluor 633 | |||||
| Alexa Fluor 647 | |||||
| Alexa Fluor 680 | |||||
| Oregon Green 488 | |||||
| Tetramethylrhodamine | |||||
| Texas Red-X | |||||
| Fluorescein | |||||
| Sampler Kit | |||||
| Wisteria floribunda lectin (WFA, WFL) | Fluorescein (FITC) | Japanese wisteria | Non-specific after neuraminidase treatment | GlcNAc | No |
Concanavalin A (Con A), a widely used fluorescent lectin marker in cell biology, selectively binds to α‑mannopyranosyl and α‑glucopyranosyl residues (Figure 1). There are several fluorescent markers of Con A and they are useful for various applications including localization of oncogene products, intracellular enzymes, viral proteins, and cytoskeletal components.
Figure 1. Osteosarcoma cells stained with Alexa Fluor 488 concanavalin A, Alexa Fluor 594 wheat germ agglutinin, and Hoechst 33342 nucleic acid stain. Fixed and permeabilized osteosarcoma cells simultaneously stained with the fluorescent lectins Alexa Fluor 488 concanavalin A (Con A) and Alexa Fluor 594 wheat germ agglutinin (WGA). Con A selectively binds a-glucopyranosyl residues, whereas WGA selectively binds sialic acid and N-acetylglucosaminyl residues. The nuclei were counterstained with blue fluorescent Hoechst 33342 nucleic acid stain. The image was acquired using bandpass filter sets appropriate for the Texas Red dye, fluorescein, and DAPI.
Wheat germ agglutinin (WGA), another widely used fluorescent lectin marker in cell biology, selectively binds to N‑acetylglucosamine and N‑acetylneuraminic acid (sialic acid) residues (Figure 2). Similar to Concanavalin A, WGA is useful for localization of oncogene products, specific intracellular enzymes, viral proteins, and cytoskeletal components.
WGA conjugates can also be used as a lectin marker for bacterial cell wall peptidoglycans, chitin, and cartilage glycosaminoglycans. WGA conjugates have been shown to stain gram-positive but not gram-negative bacteria.
Arachis hypogaea lectin (PNA), isolated from peanuts, specifically binds terminal β-galactose residues of glycoproteins (Figure 3). After neuraminidase treatment, PNA has been shown to bind human erythrocytes. Since PNA-binding sites are widespread in human tissues, the staining patterns will vary by tissue type.
For Research Use Only. Not for use in diagnostic procedures.